Skip to Content
Merck
All Photos(1)

Key Documents

11581074001

Roche

COT Human DNA

from human placenta DNA, enriched for repetitive sequences

Synonym(s):

COT Human DNA, Human DNA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106310

biological source

human placenta (DNA)

Quality Level

grade

Molecular Biology

form

solution

packaging

pkg of 500 μg

manufacturer/tradename

Roche

concentration

1 mg/mL

impurities

HCV/HBV, none detected
HIV 1/2, none detected

color

colorless

solubility

water: miscible

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Repetitive elements (IRS) present in a probe (e.g., cosmids, YACs, chromosome painting probes) generate nonspecific hybridization signals that are distributed over the whole chromosome or genome. To enable specific hybridization of the probe to the chromosomal target site (e.g., single-copy sequences or low-copy repeats) the probe must be denatured in the presence of excess unlabeled COT Human DNA. This DNA serves as a competitor. In a subsequent preannealing step, the repetitive probe elements rapidly hybridize to excess repeats in the COT Human DNA, while most of the specific probe sequences remain single-stranded and thus can be hybridized to their chromosomal targets. This technique is known as chromosomal in situ suppression (CISS) hybridization.
The COT fraction of human genomic DNA consists largely of rapidly annealing repetitive elements. These interspersed repetitive sequences (IRS) such as SINEs (small interspersed repetitive elements, e.g., Alu-elements) and LINEs (large interspersed repetitive elements, e.g., L1-elements) are distributed ubiquitously throughout the genome. COT Human DNA is prepared from human placental DNA by shearing, denaturing, and reannealing under conditions that enrich these repetitive elements.

Application

COT Human DNA is used in chromosome in situ suppression (CISS) hybridization. Cosmid or YAC probes contain repetitive elements that result in monospecific hybridization signals distributed over the entire chromosome. To enable specific hybridization to the chromosomal target site, the probe is denatured together with an excess of unlabeled COT Human DNA as a competitor. COT Human DNA can be used to suppress nonspecific hybridization to human repetitive sequences in microarray analysis, and in filter and in fluorescent in situ hybridization experiments.

Sequence

In agarose gel electrophoresis the length distribution of the COT Human DNA fragments shows a maximum in the range of 50 to 300 nucleotides.

Physical form

Solution, 1 mg/ml, 10 mM Tris-HCl, 1 mM EDTA, pH 7.4

Other Notes

For life science research only. Not for use in diagnostic procedures.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

No data available

Flash Point(C)

No data available


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

HiCTMap: Detection and Analysis of Chromosome Territory Structure and Position
by High-throughput Imaging
Ziad Jowhar
Nature (2017)
Early S-phase cell hypersensitivity to heat stress
Nadezhda V
Cell Cycle (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service