Skip to Content
Merck
All Photos(1)

Documents

11467140001

Roche

DNA, MB-grade

from fish sperm

Synonym(s):

DNA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106310

biological source

fish (sperm)

Quality Level

grade

Molecular Biology

form

solution (ready-to-use)

mol wt

40-1,000 kDa

packaging

pkg of 500 mg

manufacturer/tradename

Roche

concentration

100 μg/mL

color

light yellowopalescent

solubility

water: miscible

storage temp.

−20°C

General description

DNA, MB-grade is used for the prevention of nonspecific hybridization reactions.
DNA, MB-grade, is provided as a sonicated, denatured single-stranded DNA fragment mixture.

Application

The ready-to-use solution is directly added to the hybridization mix. It prevents nonspecific hybridization in all types of membrane (e.g., Southern and northern blot applications) or in situ DNA hybridizations. DNA, MB-grade has been used in chromatin immunoprecipitation (ChiP) assay.
Note: Addition of NaCl, sonication or shearing, and denaturing are not required.

Sequence

Chain Length 120 to 3,000 nucleotides

Physical form

Solution, 10 mg/ml, in 10 mM Tris-HCl, 10 mM NaCl, 1 mM EDTA, pH 8.0

Preparation Note

Working concentration: 100μg/ml
This fish sperm DNA should be used at a concentration of 100 μg/ml (1 : 100 dilution) in prehybridization solutions.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

No data available

Flash Point(C)

No data available


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yu Sasano et al.
Bio-protocol, 7(10), e2306-e2306 (2017-05-20)
Chromosome engineering is an important technology with applications in basic biology and biotechnology. Chromosome splitting technology called PCS (PCR-mediated Chromosome Splitting) has already been developed as a fundamental chromosome engineering technology in the budding yeast. However, the splitting efficiency of
Ryudo Ohbayashi et al.
Bio-protocol, 8(15), e2958-e2958 (2018-08-05)
Cyanobacteria are prokaryotic organisms that carry out oxygenic photosynthesis. The fresh water cyanobacterium Synechococcus elongatus PCC 7942 is a model organism for the study of photosynthesis and gene regulation, and for biotechnological applications. Besides several freshwater cyanobacteria, S. elongatus 7942
Jasmin van den Heuvel et al.
eLife, 10 (2021-07-29)
In humans and other holozoan organisms, the ribosomal protein eS30 is synthesized as a fusion protein with the ubiquitin-like protein FUBI. However, FUBI is not part of the mature 40S ribosomal subunit and cleaved off by an as-of-yet unidentified protease.
Dilip Kumar et al.
Journal of immunology (Baltimore, Md. : 1950), 182(2), 1011-1020 (2009-01-07)
The MAPKs ERK, JNK, and p38 control diverse aspects of the immune response, including regulation of cytotoxin biology in NK cells and CTL. The chemokine CCL5 is coreleased with the cytotoxins, perforin, the granzymes, and granulysin, during the lethal hit

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service