11363743001
Roche
Nutridoma™-CS
suitable for hybridoma, solution, pkg of 10 mL (50x)
Synonym(s):
serum replacements
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
sterility
sterile; 0.2 μm filtered
Quality Level
form
solution
packaging
pkg of 10 mL (50x)
manufacturer/tradename
Roche
concentration
50 ×
technique(s)
cell culture | hybridoma: suitable
impurities
mycoplasma, tested
100 EU/mL endotoxin (LAL test)
solubility
water: miscible
storage temp.
−20°C
General description
Nutridoma-CS is a serum replacement solution which helps in cell proliferation when added to a suspension. Its major component is β-D xylopyranose which has ring shaped sugar allowing the cells to grow in a suspension even without serum.
Application
Contents
Solution, 50× concentrated, Biochemically defined, filtered through 0.2μm pore size membrane, serum-free medium supplement composed of albumin, insulin, transferrin, several cytokines, a cholesterol source, and other defined organic and inorganic compounds. Nutridoma-CS contains human proteins.
Solution, 50× concentrated, Biochemically defined, filtered through 0.2μm pore size membrane, serum-free medium supplement composed of albumin, insulin, transferrin, several cytokines, a cholesterol source, and other defined organic and inorganic compounds. Nutridoma-CS contains human proteins.
Biochemically defined supplement for the replacement of fetal bovine serum (FBS) in cultures of hybridoma cells. Nutridoma-CS supports the growth of SP2/0-, P3x63Ag8.653-, and NS-1-derived hybridomas. It is specifically formulated to optimize the growth of freshly fused hybridomas during selection and cloning procedures in serum-free cell culture without feeder cells.
Nutridoma™-CS has been used as a supplement in neurobasal medium for culturing human embryonic stem cell (hESC) lines.
Unit Definition
Endotoxin (LAL): 100 EU/ml, mycoplasma tested
Physical form
Solution (50x concentrated, pH 7.4); sterile. The protein concentration is less than 1 mg/ml for 2% (1x) working concentration.
Preparation Note
Working concentration: Nutridoma-CS (50x) is diluted 1:50 (v/v) in basal medium. It is strongly suggested to use RPMI 1640. The final protein concentration is less than 1 mg/ml. The final medium should also contain L-glutamine and β-mercaptoethanol.
Other Notes
For life science research only. Not for use in diagnostic procedures.
Legal Information
Nutridoma is a trademark of Roche
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
does not flash
Flash Point(C)
does not flash
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Gastroenterology, 125(1), 89-97 (2003-07-10)
Early detection of hepatocellular carcinoma (HCC) is critical for successful treatment. However, the differential diagnosis between HCC and benign hepatic lesions is sometimes difficult and new biochemical markers for HCC are required. It has been reported that glypican-3 (GPC3) messenger
Nature biotechnology, 28(4), 361-364 (2010-03-31)
Undifferentiated human embryonic stem cells (hESCs) are currently propagated on a relatively small scale as monolayer colonies. Culture of hESCs as floating aggregates is widely used for induction of differentiation into embryoid bodies. Here we show that hESC lines can
Cellular reprogramming, 16(3), 185-195 (2014-05-09)
Four normal-karyotype human embryonic stem cell (hESC) lines were generated using the same protocol and maintained under identical conditions. Despite these precautions, gene expression patterns were found to be dissimilar among the four lines. The observed differences were typical of
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service