Skip to Content
Merck
All Photos(5)

Key Documents

ZAFS51080

Milli-Q

Milli-Q® CLX Water Purification System

Delivers up to 1600 L/day clinical laboratory reagent water (CLRW). For feed water with low chlorine levels.

Synonym(s):

Milli-Q® CLX 7080 Water Purification System (LC) 230V 50/60 Hz

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104200
eCl@ss:
33050190
NACRES:
JB.11

AC/DC input

100 V / 230 V, 50-60 Hz

Quality Level

product line

Milli-Q® CLX

packaging

pkg of 1 unit

parameter

1600 L/day max. usage
234 kg operating weight (515.9 lb)
4 L/min distribution flow rate
80 L/hr make-up flow rate

resistivity

>15 MΩ-cm, at 25°C

H × W × depth

125.5 cm (48.8 in.) × 54.3 cm (21.4 in.) × 79.7 cm (31.4 in.)

total organic carbon (TOC) residue

<30 ppb (Product water)

input

potable tap water

output

clinical laboratory reagent water (CLRW) (complies with the CLSI guideline)

Looking for similar products? Visit Product Comparison Guide

General description

The Milli-Q® CLX 7080 analyzer feed system provides an economical and high-performance water purification solution for pure water needs of up to 1600 L per day. Up to 3 systems can be placed in series to increase daily capacity. The water produced complies with the Clinical and Laboratory Standards Institute (CLSI) guideline for clinical laboratory reagent water (CLRW). This high-throughput biomedical water system supports your clinical lab′s maximum productivity by integrating state-of-the-art purification technologies and delivering an unprecedented level of system accessibility and service. MyMilli-Q Remote Care online monitoring and service capability is available on all Milli-Q® CLX systems. The Milli-Q® CLX 7080 water purification system (LC) is used when feed water has low levels of chlorine.

Application

The high-throughput Milli-Q® CLX 7080 water purification solution feeds analyzers with consistent water quality that meets CLRW standards of the CLSI. Your clinical laboratory will benefit from a high-performance and economical water purification solution that fulfills its many critical needs.

Features and Benefits

  • 24/7 real-time monitoring and secure remote-control access.
  • Automatic e-record archiving for up to two years facilitates laboratory accreditation and reaccreditation processes.
  • Patented Elix® electrodeionization (EDI) module provides consistently high-quality water with no need for resin cylinders. This reduces maintenance, running costs and analyzer downtime.
  • Unique E.R.A.® technology automatically optimizes water recovery based on feed water quality to decrease water usage costs.
  • Three built-in bactericidal UV lamps reduce system and analyzer sanitization needs to maximize lab uptime.
  • A large, user-friendly color touchscreen shows all important system information at a glance and allows for intuitive operation.
  • To ensure you always have an uninterrupted supply of pure water, these robust systems are equipped with an emergency bypass feature that is easy to set up.
  • The systems are compact and can be installed wherever it is convenient in the laboratory. They are also customizable, allowing you to adapt them to your specific requirements.
  • Available with MyMilli-Q Remote Care monitoring and service to support maximum uptime.

Components

  • The indicated price currently corresponds to the above catalog number only; it is subject to change at any time without notice.
  • This catalog number does not include consumables, accessories and other products and/or services that may be required for the installation and the correct operation of the water purification unit over time.
  • Because our customers each have specific purified water needs, we recommend that you have your purchase validated by one of our specialists. We remain at your disposal to advise you, do not hesitate to contact us!

Other Notes

Directions For Use
Organism Retention: Microorganisms
Mode of Action: Filtering and UV
Application: Clinical laboratories
Intended Use: Water purification
Instructions for Use: This item provides water filtered through 0.22 μm sterilizing-grade membranes and exposed to bactericidal UV lamp. Please refer to system equipment user guide section "System" (provided on USB key in the North American configuration version).
Storage Statement: Store in dry location
Disposal Statement: Dispose of in accordance with applicable federal, state, and local regulations.

Legal Information

E.R.A. is a registered trademark of Merck KGaA, Darmstadt, Germany
ELIX is a registered trademark of Merck KGaA, Darmstadt, Germany
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany
MyMilli-Q is a trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Critical points for laboratories to consider when assessing bacteria levels in purified water and attempting to optimize the microbiology process.

Critical points for laboratories to consider when assessing bacteria levels in purified water and attempting to optimize the microbiology process.

Critical points for laboratories to consider when assessing bacteria levels in purified water and attempting to optimize the microbiology process.

Critical points for laboratories to consider when assessing bacteria levels in purified water and attempting to optimize the microbiology process.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service