Borosilicate glass can generally be autoclaved, but we have not conducted specific tests on this item. As a result, we do not have any data or information regarding its autoclavability.
XX1514702
Millipore Base and Cap for Vacuum Filtration
47 mm, Glass frit support
Synonym(s):
Millipore filtration kit
About This Item
Recommended Products
material
borosilicate glass base
tubulated borosilicate glass cap
technique(s)
vacuum filtration: suitable
diam.
47 mm
filtration area
9.6 cm2
fitting
6 mm (1/4 in.) O.D. outlet tubulated cap sidearm for vacuum
application(s)
filtration
shipped in
ambient
Related Categories
General description
Glass Base and Cap, 47 mm, Glass frit support
These accessory products are designed to replace parts contained in Millipore glass vacuum filtration kits or provide for an alternative configuration.
This item is an accessory part for the Millipore 47 mm All-Glass Filter Holders Kits and Millipore MilliSolve Kit.
Application
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
-
Could you please confirm whether item XX1514702 is autoclavable?
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service