WMBQP0D01
Wall Mounting Bracket
Installation accessory for use with POD dispensers
Synonym(s):
Q-POD® and E-POD® Wall Mounting Bracket
About This Item
packaging
pkg of 1 unit
H × W × D
31 cm × 15 cm × 44.1 cm
compatibility
for use with E-POD®
for use with Elix® Advantage
for use with Milli-Q® IQ Element
for use with Milli-Q® IX 7003
for use with Milli-Q® IX 7005
for use with Milli-Q® IX 7010
for use with Milli-Q® IX 7015
for use with Q-POD® Element
Looking for similar products? Visit Product Comparison Guide
General description
For use with Q-POD® and E-POD® dispensers, and the Q-POD® Element unit.
Features and Benefits
- Saves lab bench space.
- Easy to install.
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service