Good to know: These filters are not white but transparent
TCTP04700
Polycarbonate Membrane Filter, 10.0 μm Pore Size
Isopore™, filter diam. 47 mm, hydrophilic
Synonym(s):
Isopore Membrane Filter
About This Item
Recommended Products
material
plain filter
polycarbonate membrane
white filter
sterility
non-sterile
feature
hydrophilic
manufacturer/tradename
Isopore™
Millipore
parameter
140 °C max. temp.
69 L/min-cm2-psi air flow rate
~900 mL/min-cm2 water flow rate at 10 psi (typical results)
filter diam.
47 mm
thickness
16 μm
gravimetric extractables
<1%
refractive index
n/D 1.6
matrix
Isopore™
pore size
10.0 μm pore size
5-20 % porosity
shipped in
ambient
Looking for similar products? Visit Product Comparison Guide
General description
Features & Benefits:
•Membrane structure retains particles on the surface, simplifying counting and analysis
•Isopore membranes do not stain, resulting in low background interference
•Non-hygroscopic, allowing for rapid drying and reduced sample analysis time
•Translucent material does not require clearing for transmitted light microscopy; also available in brown variety
Applications:
Air Monitoring, Epifluorescent Microscopy, Chemotaxis Assays
Application
- Chemotaxis
- Bioassays
- Cytology
- Air monitoring
- Microplastics analysis
Caution
Legal Information
Storage Class Code
10-13 - German Storage Class 10 to 13
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
An overview of microplastics, testing regulations, and filter selection for microplastic sample collection, preparation, and analysis.
An overview of microplastics, testing regulations, and filter selection for microplastic sample collection, preparation, and analysis.
An overview of microplastics, testing regulations, and filter selection for microplastic sample collection, preparation, and analysis.
An overview of microplastics, testing regulations, and filter selection for microplastic sample collection, preparation, and analysis.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service