Skip to Content
Merck
All Photos(1)

Key Documents

SX1248

Supelco

Sulfuric acid

OmniTrace Ultra

Synonym(s):

aa

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2SO4
CAS Number:
Molecular Weight:
98.08
Beilstein:
2037554
MDL number:
UNSPSC Code:
12352301
EC Index Number:
231-639-5
Pricing and availability is not currently available.

grade

for inorganic trace analysis

Quality Level

Agency

suitable for EPA 300

vapor density

<0.3 (25 °C, vs air)

vapor pressure

0.0001 hPa ( 20 °C)
1 mmHg ( 146 °C)

Assay

93-98%

form

liquid

concentration

≤100% in sulfuric acid

pH

0.3 (25 °C, 49 g/L in H2O)

bp

~290 °C (lit.)

mp

-20 °C

solubility

soluble (Water: Soluble (caution ! development of heat))

density

1.840 g/mL at 25 °C (lit.)

cation traces

Ag: ≤50 pg/g
Al: ≤50 pg/g
As: ≤500 pg/g
Ba: ≤10 pg/g
Be: ≤10 pg/g
Bi: ≤10 pg/g
Ca: ≤50 pg/g
Cd: ≤10 pg/g
Ce: ≤10 pg/g
Co: ≤10 pg/g
Cr: ≤10 pg/g
Cs: ≤10 pg/g
Cu: ≤10 pg/g
Dy: ≤10 pg/g
Er: ≤10 pg/g
Eu: ≤10 pg/g
Fe: ≤50 pg/g
Ga: ≤10 pg/g
Gd: ≤10 pg/g
Ge: ≤100 pg/g
Hf: ≤10 pg/g
Hg: ≤100 pg/g
Ho: ≤10 pg/g
In: ≤10 pg/g
K: ≤50 pg/g
La: ≤10 pg/g
Li: ≤10 pg/g
Lu: ≤10 pg/g
Mg: ≤50 pg/g
Mn: ≤10 pg/g
Mo: ≤10 pg/g
Na: ≤50 pg/g
Nb: ≤10 pg/g
Nd: ≤10 pg/g
Ni: ≤50 pg/g
Pb: ≤10 pg/g
Pr: ≤10 pg/g
Rb: ≤10 pg/g
Rh: ≤50 pg/g
Sb: ≤50 pg/g
Sc: ≤10 pg/g
Se: ≤500 pg/g
Sm: ≤10 pg/g
Sn: ≤50 pg/g
Sr: ≤10 pg/g
Tb: ≤10 pg/g
Te: ≤50 pg/g
Th: ≤10 pg/g
Ti: ≤50 pg/g
Tl: ≤10 pg/g
Tm: ≤10 pg/g
U: ≤10 pg/g
V: ≤10 pg/g
W: ≤10 pg/g
Y: ≤10 pg/g
Yb: ≤10 pg/g
Zn: ≤50 pg/g
Zr: ≤10 pg/g

SMILES string

OS(O)(=O)=O

shipped in

ambient

storage temp.

room temp

InChI

1S/H2O4S/c1-5(2,3)4/h(H2,1,2,3,4)

InChI key

QAOWNCQODCNURD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

OmniTrace Ultra
Product is double-distilled and packed in class 100 clean room conditions into preleached FEP bottles. Each FEP bottle is packed in an EPS innerpack and then packed into a 1x regulated carton with a Certificate of Analysis.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service