Skip to Content
Merck
All Photos(1)

Key Documents

SX0355

Supelco

Sodium Borate, Decahydrate

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

Synonym(s):

Sodium Borate, Decahydrate, Sodium Tetraborate, Dodecahydrate, Borax

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
B4Na2O7 · 10H2O
CAS Number:
Molecular Weight:
381.37
MDL number:
UNSPSC Code:
12352302
EC Index Number:
215-540-4
Assay:
99.5-105.0%
form:
crystals or granules
solubility:
49.74 g/L

Agency

meets USP/NF

Quality Level

vapor pressure

0.213 hPa ( 20 °C)

Assay

99.5-105.0%

form

crystals or granules

impurities

≤0.005% Insoluble matter

color

white

pH

9.2 (20 °C, 47 g/L in H2O)

mp

75 °C (Elimination of water of crystallisation)

solubility

49.74 g/L

anion traces

chloride (Cl-): ≤0.001%
phosphate (PO43-): ≤0.001%
sulfate (SO42-): ≤0.005%

cation traces

Ca: ≤0.005%
Fe: ≤5 ppm
heavy metals (as Pb): ≤0.001%

shipped in

ambient

storage temp.

room temp

SMILES string

[Na+].[Na+].B([O-])(OB([O-])OB=O)OB=O.O.O.O.O.O.O.O.O.O.O

InChI

1S/B4O7.2Na.10H2O/c5-1-9-3(7)11-4(8)10-2-6;;;;;;;;;;;;/h;;;10*1H2/q-2;2*+1;;;;;;;;;;

InChI key

OJPFLODMGUZMKD-UHFFFAOYSA-N

Application

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

Analysis Note

Assay99.5-105.0%Calcium (Ca)0.005% maxChloride (Cl)0.001% maxColorWhiteFormCrystals/granulesHeavy metals (as Pb)0.001% maxInsoluble matter0.005% maxIron (Fe)5ppm maxPhosphate (PO4)0.001% maxSulfate (SO4)0.005% maxpH of a 0.01 m solution at 25°C9.15-9.20

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service