Skip to Content
Merck
All Photos(2)

Documents

SVLP04700

Millipore

PVDF Membrane Filter, 5.0 μm Pore Size

Durapore®, filter diam. 47 mm, hydrophilic, white

Synonym(s):

Durapore® Membrane Filter, 5.0 µm, Hydrophilic Polyvinylidene fluoride membrane filter discs, Polyvinylidene difluoride membrane filter discs

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
40161507
eCl@ss:
32031602
NACRES:
NB.24

material

PVDF membrane
plain filter
white filter

Quality Level

sterility

non-sterile

feature

hydrophilic

manufacturer/tradename

Durapore®
Millipore

parameter

>15.4 mL/min-cm2 water flow rate
32 L/min-cm2 air flow rate
85 °C max. temp.

filter diam.

47 mm

thickness

160 μm

gravimetric extractables

0.5%

color

white

refractive index

n/D 1.42

matrix

Durapore®

pore size

5.0 μm pore size
70 % porosity

bubble point

≥0.2 bar, air with water at 23 °C

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

General description

Durapore® membranes are polyvinylidene fluoride membrane that binds less protein compared to other membranes like nylon, nitrocellulose, and polytetrafluoroethylene. It supports high flow rates, maximizes sample recovery, and displays broad chemical compatibility.
Filter Code: SVLP
Filter Type: Screen filter

Application

  • Clarifying filtration of biological solutions
  • Particle monitoring

Durapore® Membrane Filter, 5.0 μm has been used for:
  • the collection of phytoplankton from aquatic samples
  • collection of zoospores from greenhouse nursery aqueous environment samples
  • harvesting suspension cultures from various cell lines

Legal Information

Durapore is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G A Ridge et al.
Plant disease, 98(4), 551-558 (2014-04-01)
The goal of this study was to develop a procedure that could be used to evaluate the potential susceptibility of aquatic plants used in constructed wetlands to species of Phytophthora commonly found in nurseries. V8 agar plugs from actively growing
Laboratory studies on the biodegradation of organisms for estimating carbon storage potential in coastal aquatic ecosystems
Ishii Y, et al.
Coastal Engineering Journal, 1-12 (2021)
Lisa M Røst et al.
Metabolites, 10(2) (2020-02-26)
Absolute quantification of intracellular metabolite pools is a prerequisite for modeling and in-depth biological interpretation of metabolomics data. It is the final step of an elaborate metabolomics workflow, with challenges associated with all steps-from sampling to quantifying the physicochemically diverse

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service