Skip to Content
Merck
All Photos(1)

Key Documents

SLM-220-M

Sigma-Aldrich

EmbryoMax® DMEM - High Glucose, Low Bicarbonate w/o Sodium Pyruvate

The EmbryoMax DMEM -High Glucose, Low Bicarbonate w/o Sodium Pyruvate is available in a 400 mL format and may be used for routine mouse embryonic stem cell culture applications.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161702
eCl@ss:
32160801
NACRES:
NA.75
Pricing and availability is not currently available.

Quality Level

form

liquid

manufacturer/tradename

Specialty Media
EmbryoMax®

technique(s)

cell culture | stem cell: suitable

input

sample type epithelial cells
sample type induced pluripotent stem cell(s)
sample type: human embryonic stem cell(s)
sample type mesenchymal stem cell(s)
sample type pancreatic stem cell(s)
sample type cardiac stem cell(s)
sample type neural stem cell(s)
sample type: mouse embryonic stem cell(s)
sample type hematopoietic stem cell(s)

shipped in

ambient

Legal Information

EmbryoMax is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service