SCC010
ReNcell VM Immortalized Cell Kit
ReNcell VM Kit contains human neural progenitor cells, optimized maintenance media & freezing media for culture of human neural stem cells.
Synonym(s):
Human NSC Line, Human Neural Stem Cell Line, Human Neural Stem Cells
About This Item
Recommended Products
biological source
human
Quality Level
manufacturer/tradename
Chemicon®
ReNcell
technique(s)
cell culture | stem cell: suitable
input
sample type neural stem cell(s)
shipped in
liquid nitrogen
General description
Cell Line Description
Application
Stem Cell Research
Components
ReNcell NSC Maintenance Media (Cat.# SCM005)
ReNcell Neural Stem Cell Freezing Medium (Cat.# SCM007)
Quality
Other Notes
Legal Information
Disclaimer
Storage Class Code
10 - Combustible liquids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Protocols
A stem cell culture protocol to generate 3D NSC models of Alzheimer’s disease using ReNcell human neural stem cell lines.
A stem cell culture protocol to generate 3D NSC models of Alzheimer’s disease using ReNcell human neural stem cell lines.
A stem cell culture protocol to generate 3D NSC models of Alzheimer’s disease using ReNcell human neural stem cell lines.
A stem cell culture protocol to generate 3D NSC models of Alzheimer’s disease using ReNcell human neural stem cell lines.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service