Skip to Content
Merck
All Photos(2)

Key Documents

RKTX2MAG-37K

Millipore

MILLIPLEX® Rat Kidney Toxicity Magnetic Bead Panel 2 - Toxicity Multiplex Assay

The analytes available for this multiplex kit are: Albumin, AGP, ß2M, Cystatin C, EGF, Lipocalin-2/NGAL.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.84

Quality Level

species reactivity

rat

manufacturer/tradename

Milliplex®

assay range

accuracy: 70-123%
standard curve range: 0.00-2 ng/mL
(EGF)

standard curve range: 0.01-10 ng/mL
(Lipocalin-2/NGAL)

standard curve range: 0.01-8 ng/mL
(Cystatin C)

standard curve range: 0.48-350 ng/mL
(β2M)

standard curve range: 1.37-1,000 ng/mL
(AGP)

standard curve range: 10.97-8,000 ng/mL
(Albumin)

inter-assay cv: <15%
intra-assay cv: <10%
(All other analytes)

inter-assay cv: <20%
intra-assay cv: <15%
(AGP)

technique(s)

multiplexing: suitable

detection method

fluorometric (Luminex xMAP)

shipped in

wet ice

General description

MILLIPLEX® Rat Kidney Toxicity Bead Panels 1 & 2, coupled with the Luminex® xMAP®platform in a bead format, enables you to focus on discovery so you receive the advantage of ideal speed and sensitivity, allowing quantitative multiplex detection of multiple analytes simultaneously, dramatically improving productivity.

Panel Type: Toxicity

Specificity

There was no or negligible cross-reactivity between the antibodies for an analyte and any of the other analytes in this panel.

Application

  • Analytes: Alpha-1 Acid Glycoprotein (AGP), Albumin, ß2M, Cystatin C, EGF, Lipocalin-2/NGAL
  • Recommended Sample type: urine
  • Recommended Sample dilution: 1:500
  • Assay Run Time: Overnight
  • Research Category: Toxicity

Features and Benefits

Design your multiplex kit by choosing available analytes within this panel.

Packaging

Everything you need in a single kit.

Storage and Stability

Recommended storage for kit components is 2 - 8°C.

Other Notes

Sensitivity: See kit protocol for individual analytes sensitivities.

Legal Information

Luminex is a registered trademark of Luminex Corp
MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
xMAP is a registered trademark of Luminex Corp

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael J Coyne et al.
PloS one, 17(5), e0269085-e0269085 (2022-05-28)
Symmetric dimethylarginine (SDMA) is a serum biomarker of excretory renal function which consistently correlates with glomerular filtration rate (GFR) across multiple species including rats, dogs, and humans. In human and veterinary clinical settings SDMA demonstrates enhanced sensitivity for detection of
Mónica Ríos-Silva et al.
Molecules (Basel, Switzerland), 24(1) (2018-12-26)
Capsaicin is an agonist of the transient receptor potential vanilloid type 1 (TRPV1) channel, which has been related to the pathophysiology of kidney disease secondary to diabetes. This study aimed to evaluate the chronic effect of capsaicin administration on biomarkers
Iwan P Williams et al.
mAbs, 12(1), 1685349-1685349 (2019-11-27)
IgE monoclonal antibodies hold great potential for cancer therapy. Preclinical in vivo systems, particularly those in which the antibody recognizes the host species target antigen and binds to cognate Fc receptors, are often the closest approximation to human exposure and
Kidney biomarkers in MCPA-induced acute kidney injury in rats: reduced clearance enhances early biomarker performance.
Wunnapuk, K; Liu, X; Gobe, GC; Endre, ZH; Peake, PW; Grice, JE; Roberts, MS; Buckley, NA
Toxicology Letters null
Nadeeka S Udawatte et al.
Frontiers in pharmacology, 11, 511-511 (2020-05-12)
Candida albicans is the major fungal species associated with superficial mucosal infections such as oral candidiasis as well as systemic mycoses with high morbidity and mortality. On top of the rising drug resistance, currently available antifungal agents have significant adverse

Related Content

Novel kidney toxicity biomarkers expand options for acute nephrotoxicity detection. Multiplex assays measure multiple renal damage biomarkers in small sample volumes, minimizing time and costs. See how MILLIPLEX® multiplex kidney toxicity assays detected vancomycin-induced subacute nephrotoxicity in rat models.

Novel kidney toxicity biomarkers expand options for acute nephrotoxicity detection. Multiplex assays measure multiple renal damage biomarkers in small sample volumes, minimizing time and costs. See how MILLIPLEX® multiplex kidney toxicity assays detected vancomycin-induced subacute nephrotoxicity in rat models.

Novel kidney toxicity biomarkers expand options for acute nephrotoxicity detection. Multiplex assays measure multiple renal damage biomarkers in small sample volumes, minimizing time and costs. See how MILLIPLEX® multiplex kidney toxicity assays detected vancomycin-induced subacute nephrotoxicity in rat models.

Novel kidney toxicity biomarkers expand options for acute nephrotoxicity detection. Multiplex assays measure multiple renal damage biomarkers in small sample volumes, minimizing time and costs. See how MILLIPLEX® multiplex kidney toxicity assays detected vancomycin-induced subacute nephrotoxicity in rat models.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service