Skip to Content
Merck
All Photos(1)

Key Documents

MCR40027H1

Millipore

Millistak+® Pod Depth Filter

CR40, cellulose (with active carbon), filtration area 0.027 m2, hydrophilic

Synonym(s):

Millistak+ Pod Depth Filter, CR40 media series, 0.027 m2 surface area, 1/pack

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151806
eCl@ss:
32031690
NACRES:
NB.24

material

cellulose (with active carbon)
polypropylene

Quality Level

description

Activated carbon is a very versatile process aid and has a high affinity and adsorption capacity for odorous compounds and color bodies over a wide range of molecular weights

feature

hydrophilic

manufacturer/tradename

Millistak+®

parameter

2.1 bar max. differential pressure (30 psid) at 37 °C (Forward)
2.1 bar max. differential pressure (30 psid) at 37 °C (Reverse)
2.1 bar max. differential pressure (30 psid) at 4 °C (Forward)

H

14 cm (5.3 in.)

L

22 cm (8.5 in.)

filtration area

0.027 m2

impurities

<0.25 EU/mL (LAL test, Aqueous extraction)

total organic carbon (TOC) residue

1300-1700 ppb

input

sample type liquid

conductivity

4.36 – 4.57 μS/cm

filter grade

CR40

shipped in

ambient

General description

product water; post autoclave (1 cycle of 60 minutes at 123 °C) and pure water flush of 10 liters per ft²(100 L/m²) of media surface area

Specifications

Filter Grade: CR40

Preparation Note

Sterilization Method
Integrity is maintained after 2 autoclave cycles of 60 minutes @ 123 °C

Analysis Note

Total organic carbon (TOC) residue: post autoclave (1 cycle of 60 minutes at 123 °C) and pure water flush of 10 liters per ft&lt;sup&gt;2&lt;/sup&gt; (100 L/m&lt;sup&gt;2&lt;/sup&gt;) of media surface area

Legal Information

MILLISTAK+ is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service