MABT167
Anti-Transgelin/SM22a Antibody, clone 20F2.2
clone 20F2.2, from mouse
Synonym(s):
Transgelin, 22 kDa actin-binding protein, Protein WS3-10, Smooth muscle protein 22-alpha, SM22-alpha, SM22a
About This Item
Recommended Products
biological source
mouse
Quality Level
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
20F2.2, monoclonal
species reactivity
human, mouse, rat
technique(s)
immunocytochemistry: suitable
western blot: suitable
isotype
IgG1κ
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... TAGLN(6876)
General description
Immunogen
Application
Cell Structure
Cytoskeleton
Immunocytochemistry Analysis: A 1:50 dilution from a representative lot detected Transgelin/SM22a in NIH/3T3 cells.
Quality
Western Blotting Analysis: 1 µg/mL of this antibody detected Transgelin/SM22a in 10 µg of U20S cell lysate.
Target description
Physical form
Storage and Stability
Analysis Note
U20S cell lysate
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
recommended
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service