Skip to Content
Merck
All Photos(1)

Key Documents

MABN821

Sigma-Aldrich

Anti-Huntingtin Disease (HD/HTT) Antibody

mouse monoclonal, 3B5H10

Synonym(s):

Huntingtin mutants with expanded polyQ repeats, HD protein, Huntington disease protein, mHtt, PolyQ disease proteins

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43
Pricing and availability is not currently available.

Product Name

Anti-polyQ Disease Proteins Antibody, clone 3B5H10, clone 3B5H10, from mouse

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

3B5H10, monoclonal

species reactivity

human

technique(s)

electron microscopy: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
inhibition assay: suitable
western blot: suitable

isotype

IgG1

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... HTT(3064)

General description

Huntingtin (UniProt P42858; also known as HD protein, Huntington disease protein) is encoded by the HTT (also known as HD, IT15) gene (Gene ID 3064) in human. Misfolding and self-aggregation of specific proteins are a common feature of well known neurodegenerative diseases, including Huntington s disease (HD), Alzheimer s disease (AD), Parkinson s disease (PD), and amyotrophic lateral sclerosis (AML). Abnormal polyglutamine (polyQ) repeat sequence or stretch result in protein misfolding and neurodegeneration. HD, for example, is caused by polyQ expansion in the first exon-coded sequence of the causal protein Huntingtin (Htt exon 1). The expanded polyQ leads to formation of beta-sheet rich fibrillar aggregates called amyloid. Eight proteins containing polyQ tracts, but otherwise unrelated to htt, also result in protein misfolding and neurodegeneration upon polyQ expansion. Growing evidences implicate soluble oligomers or even monomers of disease-causing amyloid proteins as the more toxic species. In HD, formation of the end-stage, very large aggregated species of mutant htt (mHtt), termed an inclusion bodies (IBs), is believed to be a coping response to sequester toxic species of monomeric or small oligomeric mHtt, which are distributed diffusely throughout the neuron.

Specificity

Clone 3B5H10 targeted epitope is exposed among some of the many conformations adopted by monomeric and possibly small oligomeric polyQ species of mutant Huntingtin protein (mHtt), but disappears in higher-molecular weight aggregated forms, such as inclusion bodies (IBs). The epitope also exists in mutant forms of other polyQ-containing proteins that cause neurodegeneration, including the androgen receptor, atrophin, and ataxin-3 (Miller, J., et al. (2011). Nat. Chem. Biol. 7(12):925-934).

Immunogen

N-terminally GST-tagged human Huntingtin (Htt) fragment corresponding to exon 1-coded seqeunce containing a stretch of 66 glutamines (GST-171-Q66; Miller, J., et al. (2011). Nat. Chem. Biol. 7(12):925-934).

Application

Anti-polyQ Disease Proteins, clone 3B5H10, Cat. No. MABN821, is a highly specific mouse monoclonal antibody, that targets disease-associated polyQ proteins and has been tested in Electron Microscopy, Immunocytochemistry, Immunohistochemistry, Inhibition study, and Western Blotting.
Electron Microscopy Analysis: A representative lot immunostained transiently expressed Huntington (Htt) N-terminal fragment with 97 polyQ expansions in transfected neurons. Clone 3B5H10 labeled diffuse Htt in the cytoplasm with little staining of Htt in organelles and no staining of Htt in large inclusion bodies (IBs) (Miller, J., et al. (2011). Nat. Chem. Biol. 7(12):925-934).

Immunocytochemistry Analysis: A representative lot immunostained neurons transiently expressing Huntington (Htt) or Ataxin-3 with disease-associated polyQ expansions (Q46 to Q138), but not neurons expressing Htt with Q17 or Ataxin-3 with Q27. Clone 3B5H10 stained diffuse mutant Htt (mHtt), but not aggregated Htt in large inclusion bodies (IBs) (Miller, J., et al. (2011). Nat. Chem. Biol. 7(12):925-934).

Immunohistochemistry Analysis: A representative lot immunostained brain tissue sections from transgenic mice expressing disease-associated human mutant Huntinton (mHtt) proteins, including murine strains BACHD (full-length mHtt with Q97), R6/2 (Exon 1 fragment with ~Q150), and YAC-SCA3 (full-length mHtt with Q84). Clone 3B5H10 stained diffuse mHtt and failed to stain tissue with extensively aggregated Htt unless strong antigen retrieval by 90% formic acid treatment was performed (Miller, J., et al. (2011). Nat. Chem. Biol. 7(12):925-934).

Inhibition Analysis: A representative lot prevented the aggregation of mutant Htt (mHtt) exon 1 fragment with Q39 or Q53 and disrupted pre-aggregated mHtt oligomers and fibrils into monomer in vitro by atomic force microscopy (AFM) and dynamic light scattering (DLS) measurement (Miller, J., et al. (2011). Nat. Chem. Biol. 7(12):925-934).

Western Blotting Analysis: A representative lot detected Huntington (Htt), androgen receptor (AR), and atrophin constructs with disease-associated polyQ expansions (Q65 to Q103), but not constructs with Q19 or Q25, exogenously expressed in HEK293 and PC12 cells. Clone 3B5H10 detected monomeric and possibly small oligomeric mHtt N-terminal fragments, but not large oligomers or high molecular weight aggregates (Miller, J., et al. (2011). Nat. Chem. Biol. 7(12):925-934).

Quality

Evaluated by Western Blotting of 134Q-mtHTT and 17Q-wtHTT recombinant fragments.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected exon 1-coded N-terminal Huntington fragment with 134-glutamine/Gln (134Q) expansion, but not the corresponding normal (wild-type) fragment with 17Qs.

Target description

Variable depending on the number of polyQ. The polyQ region of Huntingtin protein (Htt) is highly polymorphic and can be expanded from 10 to 35 repeats in the normal population upto 120 or more repeats in Huntington diseased patients. 347.6 kDa calculated based on the sequence reported by UniProt P42858.

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simon Marius Galyan et al.
Scientific reports, 12(1), 19642-19642 (2022-11-18)
Currently, there are no therapies available to modify the disease progression of Huntington's disease (HD). Recent clinical trial failures of antisense oligonucleotide candidates in HD have demonstrated the need for new therapeutic approaches. Here, we developed a novel in-silico fragment
Soudabeh Imanikia et al.
Cell reports, 28(3), 581-589 (2019-07-18)
The endoplasmic reticulum unfolded protein response (UPRER) is a cellular stress response that maintains homeostasis within the secretory pathway, regulates glucose and lipid metabolism, and influences longevity. To ask whether this role in lifespan determination depends upon metabolic intermediaries, we

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service