Skip to Content
Merck
All Photos(4)

Key Documents

MABC712

Sigma-Aldrich

Anti-HAS2, clone 4E7 Antibody

clone 4E7, from mouse

Synonym(s):

Hyaluronan synthase 2, Hyaluronate synthase 2, Hyaluronic acid synthase 2, HA synthase 2, HAS2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

4E7, monoclonal

species reactivity

human

technique(s)

ELISA: suitable
immunofluorescence: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG1

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... HAS2(3037)

General description

Hyaluronan or hyaluronic acid, also known as Hyaluronan synthase 2 or Hyaluronate synthase 2 or Hyaluronic acid synthase 2 (HA synthase 2), and encoded by the gene name HAS2, is a high molecular weight unbranched polysaccharide synthesized by a wide variety of organisms from bacteria to mammals, and is a constituent of the extracellular matrix. It consists of alternating glucuronic acid and N-acetylglucosamine residues that are linked by beta-1-3 and beta-1-4 glycosidic bonds. HA is synthesized by membrane-bound synthase at the inner surface of the plasma membrane, and the chains are extruded through pore-like structures into the extracellular space. It serves a variety of functions, including space filling, lubrication of joints, and provision of a matrix through which cells can migrate. HA is actively produced during wound healing and tissue repair to provide a framework for ingrowth of blood vessels and fibroblasts. Changes in the serum concentration of HA are associated with inflammatory and degenerative arthropathies such as rheumatoid arthritis. In addition, the interaction of HA with the leukocyte receptor CD44 is important in tissue-specific homing by leukocytes, and overexpression of HA receptors has been correlated with tumor metastasis. HAS2 is a member of the newly identified vertebrate gene family encoding putative hyaluronan synthases, and its amino acid sequence shows significant homology to glycosaminoglycan synthetase (DG42) from Xenopus laevis, and human and murine hyaluronan synthase 1.

Immunogen

Purified recombinant fragment of human HAS2 expressed in E. Coli

Application

Anti-HAS2, clone 4E7 Antibody is an antibody against HAS2 for use in western blotting, IHC, immunofluorescence & ELISA.
Immunohistochemistry Analysis: A 1:200-1,000 dilution from a representative lot detected HAS2 in ovarian cancer tissue.

Immunofluorescence Analysis: A 1;100-500 dilution from a representative lot detected HAS2 in HeLa cells.

ELISA: A representative lot detected control antigen (100 ng) and antigen (10 ng, 50 ng, 100 ng) in ELISA (Direct).
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional

Quality

Evaluated by Western Blotting in human plasma and Jurkat lysates.

Western Blotting Analysis: A 1:500-2,000 dilution of this antibody detected HAS2 in human plasma and Jurkat lysates.

Target description

~55 kDa observed. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal in PBS containing up to 0.1% sodium azide and 0.5% protein stabilizer.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service