Skip to Content
Merck
All Photos(1)

Key Documents

MABC1140

Sigma-Aldrich

Anti-Mesothelin Antibody, clone MN-1

clone MN-1, from mouse

Synonym(s):

CAK1 antigen, Pre-pro-megakaryocyte-potentiating factor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43
Pricing and availability is not currently available.

biological source

mouse

antibody form

purified antibody

antibody product type

primary antibodies

clone

MN-1, monoclonal

species reactivity

human

packaging

antibody small pack of 25 μg

technique(s)

ELISA: suitable
flow cytometry: suitable
immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... MSLN(10232)

General description

Mesothelin (UniProt: Q13421; also known as CAK1 antigen, Pre-pro-megakaryocyte-potentiating factor, MSLN) is encoded by the MSLN (also known as MPF) gene (Gene ID: 10232) in human. Mesothelin is a glycophosphatidylinositol (GPI)-linked cell surface protein that normally expressed in lung and at lower levels in heart, placenta, and kidney. It is highly expressed in several types of malignant tumors and is associated with increased tumor aggressiveness and poor clinical outcome. Antibodies against mesothelin have been detected in patients with mesothelioma and ovarian cancer. Mesothelin plays a role in cellular adhesion. Mesothelin is synthesized with a signal peptide (aa 1-36) and a propeptide (aa 607-630) that are cleaved to produce mature mesothelin, which can be cleaved further by furin-like convertase to generate megakaryocyte-potentiating factor (aa 37-206) and the cleaved form of mesothelin (aa 296-606). Four isoforms of mesothelin have been described that are produced by alternative splicing. (Ref.: Inaguma, S., et al. (2017). Oncotarget. 8(16); 26744-26754; Onda, M., et al. (2005). Clin. Cancer Res, 11(16); 5840-5846).

Specificity

Clone MN-1 is a mouse monoclonal antibody, generated in mesothelin-deficient mice, which specifically detects human Mesothelin.

Immunogen

Plasmid cDNA corresponding to the mesothelin extracellular domain followed by a single boost with a recombinant human mesothelin-Fc fusion protein.

Application

Anti-Mesothelin, clone MN-1 Antibody, Cat. No. MABC1140, is a mouse monoclonal antibody that detects Mesothelin and has been tested for use in ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), and Western Blotting.
Immunohistochemistry (Paraffin) Analysis: A 1:50 dilution from a representative lot detected Mesothelin in human uterus tissue sections.

Flow Cytometry Analysis: A representative lot detected Mesothelin in Flow Cytometry applications (Onda, M., et. al. (2005). Clin Cancer Res. 11(16):5840-6).

ELISA Analysis: A representative lot detected Mesothelin in ELISA applications (Onda, M., et. al. (2005). Clin Cancer Res. 11(16):5840-6).

Western Blotting Analysis: A representative lot detected Mesothelin in Western Blotting applications (Onda, M., et. al. (2005). Clin Cancer Res. 11(16):5840-6).

Immunohistochemistry (Paraffin) Analysis: A representative lot detected Mesothelin in Immunohistochemistry applications (Onda, M., et. al. (2005). Clin Cancer Res. 11(16):5840-6; Inaquma, S., et. al. (2017). Oncotarget. 8(16):26744-26754).
Research Category
Apoptosis & Cancer

Quality

Evaluated by Immunohistochemistry (Paraffin) in human fallopian tube tissue sections.

Immunohistochemistry (Paraffin) Analysis: A 1:250 dilution of this antibody detected Mesothelin in human fallopian tube tissue sections.

Target description

68.99 kDa calculated.

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service