Skip to Content
Merck
All Photos(1)

Key Documents

MAB8782

Sigma-Aldrich

Anti-Parainfluenza 4 Antibody, clone 531-1G

ascites fluid, clone 531-1G, Chemicon®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
531-1G, monoclonal
application:
ELISA
IF
IP
WB
species reactivity:
human
technique(s):
ELISA: suitable
immunofluorescence: suitable
immunoprecipitation (IP): suitable
western blot: suitable
citations:
1

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

531-1G, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunofluorescence: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

shipped in

dry ice

Specificity

Specific for Parainfluenza 4A and 4B. No cross-reactivity to Influenza A, Parainfluenza 1, 2 or 3, Adenovirus, RSV, or Mumps.

Immunogen

Fusion protein of Parainfluenza 4A, strain M-25

Application

Indirect immunofluorescence, Enzyme immunoassay, Western blotting.

Optimal working dilutions must be determined by the end user.
Research Category
Infectious Diseases
Research Sub Category
Infectious Diseases - Viral
This Anti-Parainfluenza 4 Antibody, clone 531-1G is validated for use in ELISA, IF, IP, WB for the detection of Parainfluenza 4.

Physical form

Ascites fluid, with 0.1% sodium azide.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 6 months after date of receipt. Avoid repeated freeze/thaw cycles.

WARNINGS / PRECAUTIONS:

Sodium Azide may react with lead or copper plumbing to form explosive metal azides. When disposing of Sodium Azide, flush plumbing with a large volume of water to prevent build-up.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Manuel V Villaran et al.
Influenza and other respiratory viruses, 8(2), 217-227 (2013-11-30)
Human parainfluenza viruses (HPIVs) are common viral causes of community-acquired pneumonia, particularly in children. The four types of HPIV have world-wide distribution; however, limited information exists about the epidemiological profile of HPIV in Latin-America. Provide epidemiologic and phylogenetic information about

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service