Skip to Content
Merck
All Photos(1)

Key Documents

MAB1285FX

Sigma-Aldrich

Anti-HLA-B27 Antibody, clone HLA.ABC.m3, FITC conjugated

clone HLA.ABC.m3, Chemicon®, from mouse

Synonym(s):

Anti-AS, Anti-HLAB

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43
clone:
HLA.ABC.m3, monoclonal
application:
FACS
species reactivity:
human
technique(s):
flow cytometry: suitable
citations:
1

biological source

mouse

Quality Level

100
300

conjugate

FITC conjugate

antibody form

purified antibody

antibody product type

primary antibodies

clone

HLA.ABC.m3, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

flow cytometry: suitable

isotype

IgG2a

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... HLA-B(3106)

General description

BIOCHEMISTRY: pI = 8.0
The previously assigned protein identifier P03989 has been merged into P01889. Full details can be found on the UniProt database.

Specificity

This antibody binds strongly to the HLA-B27 antigen. It also binds less strongly to the HLA-B7 antigen. Affinity studies by Scatchard analysis showed that the antibody has a higher affinity for HLA-B27 (9.7x108M-1) than for HLA-B7 (9.5x107M-).

Immunogen

Immune complex precipitated from HLA-B27 positive cell line (Bordin) by anti HLA antibody and staphylococcal Protein A.

Application

Detect HLA-B27 using this Anti-HLA-B27 Antibody, clone HLA.ABC.m3, FITC conjugated validated for use in FC.
Research Category
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology
This antibody is useful in the investigation of HLA-B27 in diseases such as ankylosing spondylitis and anterior uveitis.

SUGGESTED USAGE DILUTION

Flow cytometry and immunofluorescence 1:15 to 1:60

Dilute with isotonic buffer. Use 50 μl of diluted antibody per 100 μl of whole blood, or 1 x 10E6 peripheral blood mononuclear cells (PBMC) in 100 μl buffer.

Physical form

Purified from mouse ascitic fluid. Fluorescein conjugated. The antibody is supplied in PBS 0.01M, Sodium Azide 0.1%, pH 7.1, 0.2% BSA. The characteristics of each lot are tested by flow cytometry.

Storage and Stability

Store at 2 to 8°C. DO NOT FREEZE. Protect from light.

WARNING: The monoclonal reagent solution contains 0.1% sodium azide as a preservative. Due to potential hazards arising from the build up of this material in pipes, spent reagent should be disposed of with liberal volumes of water.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Flow cytometry dye selection tips match fluorophores to flow cytometer configurations, enhancing panel performance.

Flow cytometry dye selection tips match fluorophores to flow cytometer configurations, enhancing panel performance.

Troubleshooting guide offers solutions for common flow cytometry problems, ensuring improved analysis performance.

Troubleshooting guide offers solutions for common flow cytometry problems, ensuring improved analysis performance.

See All

Protocols

Learn key steps in flow cytometry protocols to make your next flow cytometry experiment run with ease.

Explore our flow cytometry guide to uncover flow cytometry basics, traditional flow cytometer components, key flow cytometry protocol steps, and proper controls.

Explore our flow cytometry guide to uncover flow cytometry basics, traditional flow cytometer components, key flow cytometry protocol steps, and proper controls.

Learn key steps in flow cytometry protocols to make your next flow cytometry experiment run with ease.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service