M00PMCB24
m-ColiBlue24 Broth
ready-to-use, ampule of 2 mL, suitable for microbiology, for Escherichia spp., for coliforms
Synonym(s):
EC Broth, Coliform Broth
About This Item
Recommended Products
form
liquid
Quality Level
shelf life
12 mo.
feature
ready-to-use
packaging
ampule of 2 mL
pack of 50 tests
color
blue
input
sample type sport drink
sample type cider
sample type beer
sample type cosmetics
sample type food(s)
sample type wine
sample type bottled water
sample type beverage(s)
sample type soft drink
pH
7.0±0.2 (25.0 °C)
application(s)
bioburden testing
clinical
cosmetics
environmental
food and beverages
industrial qc
water monitoring
microbiology
compatibility
for use with MF-method
for use with Microfil®
for use with MicropreSure®
for use with Milliflex®
shipped in
wet ice
storage temp.
2-8°C
suitability
Escherichia spp.
coliforms
General description
Application
Features and Benefits
- Ready-to-use Liquid ampoule media solve numerous problems in microbiology laboratories where time, equipment, and trained personnel are often in short supply.
- They are available in convenient, single-test 2 mL plastic ampoules, eliminating the manipulation of glass ampoules and avoiding the waste and spoilage associated with bulk packaged media.
Specifications
Analysis Note
E. cloacae (ATCC 23355)
E. coli (ATCC 25922)
K. pneumoniae (ATCC 13883)
P. aeruginosa (ATCC 27853)
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Sens. 1
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service