Skip to Content
Merck
All Photos(2)

Documents

HNDG1MAG-36K

Millipore

MILLIPLEX® Human Neurodegenerative Disease Magnetic Bead Panel 1 - Neuroscience Multiplex Assay

The analytes available for this multiplex kit are: α2-Macroglobulin, Apo Al, Apo CIII, Apo E, Complement C3 and Complement Factor H.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.84

Quality Level

species reactivity

human

manufacturer/tradename

Milliplex®

assay range

accuracy: 105%
(Complement Factor H)

accuracy: 110%
(Apo E)

accuracy: 75%
(Complement C3)

intra-assay cv: <10
standard curve range: 0.01-40 ng/mL
(Apo CIII)

standard curve range: 0.05-200 ng/mL
(Apo E, Complement C3)

standard curve range: 0.3-1,000 ng/mL
(Complement Factor H)

standard curve range: 0.5-2,000 ng/mL
(α-2-Macroglobulin)

standard curve range: 0.7-3,000 ng/mL
(Apo A1)

technique(s)

multiplexing: suitable

detection method

fluorometric (Luminex xMAP)

shipped in

wet ice

General description

Neurodegenerative disease is a condition characterized by the deterioration of neurons or their myelin sheath over time in the brain and/or spinal cord. These neurons are responsible for such everyday activities as processing sensory information, making decisions, and controlling movement. Because these cells are not easily regenerated, excessive cumulative damage can lead to age-related diseases such as Alzheimer′s and Parkinson′s disease, as well as other conditions such as amyotrophic lateral sclerosis (ALS) and epilepsy. These disorders are devastating and expensive, both on a personal and global level, and as population demographics continue to change, a therapeutic solution is critical. Consequently, research is underway to identify biomarkers that will help scientists not only understand the pathogenesis of neurodegenerative disease, but also identify people with these disorders before the onset of symptoms and potentially provide new therapeutic tools.
The “MILLIPLEX” Human Neurodegenerative Bead Panel 1 (HNDG1MAG-36K) is to be used for the simultaneous quantification of the following 7 analytes in any combination: α-2-Macroglobulin, Apo A1, Apo CIII, Apo E, Complement C3 and Complement Factor H. This kit may be used for the analysis of all above analytes in human serum, plasma, and cerebrospinal fluid samples.

Panel Type: Neuroscience

Specificity

Cross Reactivty
Cross-reactivity between the antibodies and any of the other analytes in this panel is non-detectable or negligible.

Application

  • Analytes: : α2-Macroglobulin, Apo Al, Apo CIII, Apo E, Complement C3, Complement Factor H
  • Recommended Sample type: serum, plasma and CSF
  • Recommended Sample dilution: 1:40,000 diluted serum/plasma or 1:400 diluted CSF per well
  • Assay Run Time: One day
  • Research Category: Neuroscience

Features and Benefits

Design your multiplex kit by choosing available analytes within this panel.

Packaging

Everything you need in a single kit.

Storage and Stability

Recommended storage for kit components is 2 - 8°C.

Other Notes

Sensitivity: See protocol for sensitivities of individual analytes

Legal Information

MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Sens. 1 - STOT RE 2

Target Organs

Respiratory Tract

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

William T Hu et al.
Acta neuropathologica communications, 4, 14-14 (2016-02-19)
CSF levels of established Alzheimer's disease (AD) biomarkers remain stable despite disease progression, and non-amyloid non-tau biomarkers have the potential of informing disease stage and progression. We previously identified complement 3 (C3) to be decreased in AD dementia, but this
Cerebrospinal fluid complement activation in patients with pneumococcal and meningococcal meningitis.
Mook-Kanamori, BB; Brouwer, MC; Geldhoff, M; Ende, Av; van de Beek, D
The Journal of Infection null
Abdul Hye et al.
Alzheimer's & dementia : the journal of the Alzheimer's Association, 10(6), 799-807 (2014-07-12)
The study aimed to validate previously discovered plasma biomarkers associated with AD, using a design based on imaging measures as surrogate for disease severity and assess their prognostic value in predicting conversion to dementia. Three multicenter cohorts of cognitively healthy
Maria H Pham et al.
Materials (Basel, Switzerland), 13(3) (2020-02-08)
Immediately after dental implant insertion, blood will be in direct contact and interact with the implant surface and activates inflammatory responses and complement cascades within seconds. The aim of the present study was to determine the ability of fluoride-modified titanium
Zaohuo Cheng et al.
Frontiers in aging neuroscience, 10, 414-414 (2019-01-09)
It is well known that Alzheimer's disease (AD) is one of the most common progressive neurodegenerative diseases; it begins gradually, and therefore no effective medicine is administered in the beginning. Thus, early diagnosis and prevention of AD are crucial. The

Related Content

Multiplex detection of Alzheimer's biomarkers with MILLIPLEX® Human Amyloid Beta and Tau Magnetic Bead Panel aids research.

Multiplex detection of Alzheimer's biomarkers with MILLIPLEX® Human Amyloid Beta and Tau Magnetic Bead Panel aids research.

Multiplex detection of Alzheimer's biomarkers with MILLIPLEX® Human Amyloid Beta and Tau Magnetic Bead Panel aids research.

Multiplex detection of Alzheimer's biomarkers with MILLIPLEX® Human Amyloid Beta and Tau Magnetic Bead Panel aids research.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service