Skip to Content
Merck
All Photos(1)

Key Documents

FX0245

Supelco

Ferrous Ammonium Sulfate, Hexahydrate

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

Synonym(s):

Ferrous Ammonium Sulfate, Hexahydrate, Ammonium Iron(II) Sulfate, Hexahydrate, Ammonium Iron (II) Sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Fe(NH4)2(SO4)2 · 6H2O
CAS Number:
MDL number:
UNSPSC Code:
12352302
EC Index Number:
233-151-8
Assay:
98.5-101.5%
form:
crystals
solubility:
269 g/L
Pricing and availability is not currently available.

Quality Level

Assay

98.5-101.5%

form

crystals

impurities

≤0.01% Insoluble matter

color

blue-green

pH

3-5 (20 °C, 50 g/L in H2O)

mp

39-41 °C (Elimination of water of crystallisation)

solubility

269 g/L

anion traces

phosphate (PO43-): ≤0.003%

cation traces

Ca: ≤0.005%
Cu: ≤0.003%
Fe3+: ≤0.01%
K: ≤0.002%
Mg: ≤0.002%
Mn: ≤0.01%
Na: ≤0.02%
Zn: ≤0.003%

shipped in

ambient

storage temp.

room temp

SMILES string

[Fe+3].[S](=O)(=O)([O-])[O-].[S](=O)(=O)([O-])[O-].[N+H4].[N+H4].O.O.O.O.O.O

InChI

1S/Fe.2H3N.2H2O4S.6H2O/c;;;2*1-5(2,3)4;;;;;;/h;2*1H3;2*(H2,1,2,3,4);6*1H2/q+3;;;;;;;;;;/p-2

InChI key

SHHFGSWBFZVUNE-UHFFFAOYSA-L

Related Categories

Analysis Note

Assay: 98.5-101.5%
Calcium (Ca): 0.005% max
Color: Blue-green (no yellow)
Copper (Cu): 0.003% max
Ferric iron (Fe+++): 0.01% max
Form: Crystals
Insoluble matter: 0.01% max
Magnesium (Mg): 0.002% max
Manganese (Mn): 0.01% max
Phosphate (PO4): 0.003% max
Potassium (K): 0.002% max
Sodium (Na): 0.02% max
Zinc (Zn): 0.003% max

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service