Skip to Content
Merck
All Photos(2)

Key Documents

EM3234

Supelco

Water

Deionized Distilled, ASTM Type II

Synonym(s):

Water

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2O
CAS Number:
Molecular Weight:
18.02
Beilstein:
2050024
MDL number:
UNSPSC Code:
50202301
bp:
100 °C (lit.)
vapor pressure:
23 hPa ( 20 °C)
Pricing and availability is not currently available.

Product Name

Water, Deionized Distilled, ASTM Type II, Distilled, ASTM Type II

vapor pressure

23 hPa ( 20 °C)

Quality Level

description

TOC <= 50g/L (at the time of manufacture)

form

liquid

shelf life

18 mo. (from manufacture date)

dilution

(for analytical testing)

impurities

≤0.01 ppm Heavy metals (as Pb)
≤0.4 ppm Chloride
≤0.4 ppm Nitrate
≤1.0 ppm Phosphate
≤1.0 ppm Sulfate
≤20 ppm Total solids
≤3 ppb Silica (SiO2)
≤5 ppb Chloride (Cl)

color

APHA: ≤10

refractive index

n20/D 1.34 (lit.)

conductivity

≤1 S/cm at 25 °C

bp

100 °C (lit.)

mp

0 °C (lit.)

density

1.000 g/mL at 3.98 °C (lit.)

anion traces

chloride (Cl-): ≤5 ppb

cation traces

Fe: ≤0.3 ppm
Na: ≤5 ppb

shipped in

ambient

storage temp.

no temp limit

SMILES string

O

InChI

1S/H2O/h1H2

InChI key

XLYOFNOQVPJJNP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Distilled, ASTM Type II

Analysis Note

Expiration date: 18 months from mfg date
Chloride (Cl): 5ppb max
Color (APHA): 10 max
Conductivity at 25°C: 1.0S/cm max
Form: Clear liquid
Heavy metals (as Pb): 0.01ppm max
Ion chromatography, Chloride: 0.4ppm max
Ion chromatography, Nitrate: 0.4ppm max
Ion chromatography, Phosphate: 1.0ppm max
Ion chromatography, Sulfate: 1.0ppm max
Iron (Fe): 0.3ppm max
Microbial limits (at time of manufacture): 10CFU/mL max
Resistivity at 25°C: 1.0MOhm❞cm min
Silica (SiO2): 3ppb max
Sodium (Na): 5ppb max
Substances reducing permanganate: To pass test
Total organic carbon (at time of manufacture): 50g/L max
Total solids: 20ppm max

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

No data available

Flash Point(C)

No data available


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 4

1 of 4

Questions

1–3 of 3 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  3. Does this water meet the ASTM Intl. specification for Type 2 subtype A

    1 answer
    1. This product meets the ASTM International specifications for Type II. The specific subtype is not determined.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service