Skip to Content
Merck
All Photos(1)

Documents

DX1730

Supelco

N,N-Dimethylformamide

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

Synonym(s):

Formic Acid Dimethylamide, DMF, DMF, NSC 5356

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCON(CH3)2
CAS Number:
Molecular Weight:
73.09
Beilstein:
605365
MDL number:
UNSPSC Code:
12352111
EC Index Number:
200-679-5

vapor density

2.5 (vs air)

Quality Level

vapor pressure

2.7 mmHg ( 20 °C)

Assay

≥99.8% (GC)

form

liquid

autoignition temp.

833 °F

shelf life

24 mo. (from mfg date)

expl. lim.

15.2 %

impurities

≤0.0005 meq/g Titrable acid
≤0.003 meq/g Titrable base
≤0.15% Water (H2O)
≤10 ppm Free amine (as dimethylamine)

evapn. residue

≤0.005%

color

clearAPHA: ≤15

refractive index

n20/D 1.430 (lit.)

pH

7 (20 °C, 200 g/L in H2O)

bp

153 °C (lit.)

mp

−61 °C (lit.)

transition temp

flash point 57.5 °C

density

0.944 g/mL (lit.)

shipped in

ambient

storage temp.

no temp limit

SMILES string

[H]C(=O)N(C)C

InChI

1S/C3H7NO/c1-4(2)3-5/h3H,1-2H3

InChI key

ZMXDDKWLCZADIW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Solvent for many hydrophobic organic compounds.

Analysis Note

Expiration date: Exp 24 months from mfg date
Appearance: Clear
Assay (GC): 99.8% min
Color (APHA): 15 max
Evaporation residue: 0.005% max
Form: Liquid
Free amine (as dimethylamine): 10ppm max
Titrable acid: 0.0005meq/g max
Titrable base: 0.003meq/g max
Water (H2O): 0.15% max

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Flam. Liq. 3 - Repr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

135.5 °F - closed cup

Flash Point(C)

57.5 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service