Skip to Content
Merck
All Photos(1)

Key Documents

CBL415P

Sigma-Aldrich

Anti-Thy-1 Antibody, clone F15-42-1, Phycoerythrin conjugated

clone F15-42-1, Chemicon®, from mouse

Synonym(s):

CD90

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
conjugate:
PE
application:
FACS
clone:
F15-42-1, monoclonal
species reactivity:
human
citations:
1
technique(s):
flow cytometry: suitable

biological source

mouse

Quality Level

conjugate

PE

antibody form

purified antibody

antibody product type

primary antibodies

clone

F15-42-1, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

flow cytometry: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... THY1(7070)

Specificity

Thy-1 molecule is heavily glycosylated membrane bound protein of 18 kDa

Application

Detect Thy-1 using this Anti-Thy-1 Antibody, clone F15-42-1, Phycoerythrin conjugated validated for use in FC.
Research Category
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology
Studies of frozen, unfixed, tissue sections of Thy.1 antigen, notably in studies of neuroblastoma, leukaemia phenotyping and carcinoma of the breast

Studies of renal corticol fibroblasts in progressive renal disease

Epitope is resistant to trypsin treatment used to release adherent cells from surfaces. Antibody not tested in fixed tissue.

Optimal working dilutions must be determined by the end user.

Linkage

Replaces: CBL1323P

Physical form

The conjugate is supplied as a 100 test vial in phosphate buffered saline containing 10mM sodium azide and 1mg/ml bovine serum albumin. For flow cytometry we recommend using 10μL of conjugate per test.

Storage and Stability

Store at +4°C protected from light. DO NOT FREEZE. For long term use and storage aliquot conjugate into small volumes and store at +4°C.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gabriel Kolle et al.
Stem cells (Dayton, Ohio), 27(10), 2446-2456 (2009-08-04)
Surface marker expression forms the basis for characterization and isolation of human embryonic stem cells (hESCs). Currently, there are few well-defined protein epitopes that definitively mark hESCs. Here we combine immunotranscriptional profiling of hESC lines with membrane-polysome translation state array

Articles

Frequently asked questions about mesenchymal stem cells including MSC derivation, expansion, differentiation and allogenic stem cell therapy.

Frequently asked questions about mesenchymal stem cells including MSC derivation, expansion, differentiation and allogenic stem cell therapy.

Frequently asked questions about mesenchymal stem cells including MSC derivation, expansion, differentiation and allogenic stem cell therapy.

Frequently asked questions about mesenchymal stem cells including MSC derivation, expansion, differentiation and allogenic stem cell therapy.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service