Skip to Content
Merck
All Photos(1)

Key Documents

AP106C

Sigma-Aldrich

Rabbit Anti-Goat IgG Antibody, Cy3 conjugate

Chemicon®, from rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.46

biological source

rabbit

Quality Level

conjugate

CY3 conjugate

antibody form

F(ab′)2 fragment of affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

species reactivity

goat

manufacturer/tradename

Chemicon®

technique(s)

immunofluorescence: suitable

shipped in

wet ice

target post-translational modification

unmodified

Application

Research Category
Secondary & Control Antibodies
Research Sub Category
Whole Immunoglobulin Secondary Antibodies
This Rabbit anti-Goat IgG Antibody, Cy3 conjugate is validated for use in IF for the detection of Goat IgG.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ayako Hakucho et al.
Hepatology research : the official journal of the Japan Society of Hepatology, 44(5), 560-570 (2013-04-24)
Oxidative stress is a major pathway mediating ethanol hepatotoxicity and liver injury. We previously found that carvedilol, which can block the sympathetic nervous system via β1-, β2- and α1-adrenoreceptors, modifies ethanol-induced production of lipogenesis- and fibrogenesis-related mediators from hepatic stellate
Vivian W Tang
Biology direct, 1, 37-37 (2006-12-13)
Zonula occludens, also known as the tight junction, is a specialized cell-cell interaction characterized by membrane "kisses" between epithelial cells. A cytoplasmic plaque of approximately 100 nm corresponding to a meshwork of densely packed proteins underlies the tight junction membrane
Daniela Planska et al.
Anticancer research, 38(8), 4629-4636 (2018-08-01)
Melanoma is a cancer disease with increasing incidence in the Caucasian population. It is often accompanied by spontaneous regression (SR), probably due to high immunogenicity. Understanding of this phenomenon could allow its induction in clinical practice, but detailed study in
Avtar S Meena et al.
Cell reports, 39(11), 110937-110937 (2022-06-16)
Intestinal epithelial tight junction disruption is a primary contributing factor in alcohol-associated endotoxemia, systemic inflammation, and multiple organ damage. Ethanol and acetaldehyde disrupt tight junctions by elevating intracellular Ca2+. Here we identify TRPV6, a Ca2+-permeable channel, as responsible for alcohol-induced
Yiyu Deng et al.
PloS one, 9(1), e87420-e87420 (2014-02-06)
Hypoxic exposure in the perinatal period causes periventricular white matter damage (PWMD), a condition associated with myelination abnormalities. Under hypoxic conditions, glial cells were activated and released a large number of inflammatory mediators in the PWM in neonatal brain, which

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service