Skip to Content
Merck
All Photos(1)

Documents

AG374

Sigma-Aldrich

Metabotropic Glutamate Receptor Peptide

control for AB5675, liquid

Synonym(s):

mGluR5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.42

product name

Metabotropic Glutamate Receptor 5, control peptide for AB5675,

biological source

rat

Quality Level

form

liquid

manufacturer/tradename

Chemicon®

concentration

1 mg/mL

technique(s)

cell based assay: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

Gene Information

rat ... Grm5(24418)

General description

Metabotropic Glutamate Receptor 5 (mGluR5) control peptide.

Application

Antibody blocking (AB5675)

Optimal working concentration must be determined by the end user.
Research Category
Neuroscience
Research Sub Category
Neurotransmitters & Receptors

Physical form

Liquid.

Storage and Stability

Store at -20°C in undiluted aliquots for up to 6 months after date of receipt. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Research design for the project information for analysis of clinical care in the Academic Medical Center, Amsterdam.
H Prins,J H Zwetsloot-Schonk,H Buller
Medinfo. MEDINFO null
Expression of metabotropic glutamate receptor mGluR5 in human dental pulp.
Yun Sook Kim,Young Jae Kim,Sang Kyoo Paik,Yi Sul Cho,Tae Geon Kwon,Dong Kuk Ahn et al.
Journal of Endodontics null
A H Iyo et al.
Neuroscience, 169(4), 1567-1574 (2010-07-06)
Several lines of evidence suggest a dysfunctional glutamate system in major depressive disorder (MDD). Recently, we reported reduced levels of metabotropic glutamate receptor subtype 5 (mGluR5) in postmortem brains in MDD, however the neurobiological mechanisms that induce these abnormalities are

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service