Skip to Content
Merck
All Photos(3)

Key Documents

ABT1485-AF488

Sigma-Aldrich

Anti-Actin Antibody, Alexa Fluor 488 Conjugate

from rabbit, ALEXA FLUOR 488

Synonym(s):

Actin, alpha skeletal muscle, Alpha-actin-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.45
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

conjugate

ALEXA FLUOR 488

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, mouse

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
immunofluorescence: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ACTA1(58)

General description

Actins are globular multi-functional proteins that serve as the basic building blocks of cytoskeletal microfilaments and are among the most conserved eukaryotic proteins. Six actin types exist, skeletal muscle alpha-actin (ACTA1), smooth muscle alpha-actin (ACTA2), cytoplasmic beta-actin (ACTB), cardiac muscle alpha-actin (ACTC1), cytoplasmic gamma-actin (ACTG1), and smooth muscle gamma-actin (ACTG2, a.k.a. ACTA3). Although actins show more than 90% overall sequence homology, isotypes do show spatial, temporal, and tissue-specific expresson patterns and only 50-60% homology is found in their 18 N-terminal residues. Cytoplasmic β and γ-actins are thought to be present in all cells, while the other four actin isoforms are typically found in specific adult muscle tissue types. Actins exist in a variety of structural states, depending on the specific ionic conditions or the interaction with ligand proteins. The oligomeric and polymeric forms that actin molecules assume are dependent on the distinct conformations they adopt. In additon to serving as the basic building blocks of cytoskeletal microfilaments, actins are involved in multiple nuclear functions and adopt specific conformations that are important for their association with nuclear actin-binding proteins.

Specificity

Broad species reactivity expected due to 100% sequence homology, including Bovine, Horse, Non-Human Primate, Porcine, Rabbit, and Rat.
This Alexa Fluor 488 conjugate was prepared from purified rabbit polyclonal antibody (Cat. No. ABT1485) raised against skeletal muscle alpha-actin (ACTA1) N-terminal end sequence that shares 80% sequence identity in the corresponding region with cardiac muscle alpha-actin (ACTC1), smooth muscle alpha-actin (ACTA2), and cytoplasmic gamma-actin (ACTG1). Immunogen sequence is not found in smooth muscle gamma-actin (ACTG2, a.k.a. ACTA3) and cytoplasmic beta-actin (ACTB).

Immunogen

Epitope: N-terminus
KLH-conjugated linear peptide corresponding to the N-terminal end sequence of mature skeletal muscle alpha-actin (ACTA1).

Application

Flow Cytometry Analysis: 2 µL from a representative lot detected actin in one million HeLa cells.

Immunofluorescence Analysis: A 1:150 dilution from a representative lot detected actin in human heart and skin muscle tissue sections.

The unconjugated antibody (Cat. No. ABT1485) and Alexa Fluor 647-conjugated antibody (Cat. No. ABT1485-AF647) are also available for flow cytometry, immunocytochemistry, immunofluorescence, immunohistochemistry and Western blotting applications.
Research Category
Cell Structure
This rabbit polyclonal Anti-Actin, Alexa Fluor 488 Conjugate, Cat. No. ABT1485-AF488 is validated for use in Flow Cytometry, Immunocytochemistry and Immunofluorescence applications for the detection of Actin.

Quality

Evaluated by Immunocytochemistry in HeLa cells.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected both cytoplasmic and nuclear actin immunoreactivity in HeLa cells.

Target description

~42 kDa observed.

Physical form

Affinity purified.
Purified rabbit polyclonal antibody conjugate in PBS with 1.5% BSA and 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Legal Information

ALEXA FLUOR is a trademark of Life Technologies

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service