Skip to Content
Merck
All Photos(8)

Key Documents

AB7356

Sigma-Aldrich

Anti-von Willebrand Factor Antibody

Chemicon®, from rabbit

Synonym(s):

Factor VIII Related Antigen

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, rat, mouse

packaging

antibody small pack of 25 μg

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunohistochemistry: suitable (paraffin)

NCBI accession no.

UniProt accession no.

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

General description

This Anti-von Willebrand Factor Antibody is validated for use in ELISA, Immunohistochemistry (IHC) for the detection of von Willebrand Factor.
von Willebrand factor (UniProt: P04275; also known as vWF) is encoded by the VWF (also known as F8VWF) gene (Gene ID:7450) in human. vWF is a multimeric plasma glycoprotein that is synthesized by endothelial cells and plays an important in the maintenance of hemostasis. It is synthesized with a signal peptide (aa 1-22), which is subsequently cleaved off in the mature form. vWF is known to promote adhesion of platelets to the sites of vascular injury by forming a molecular bridge between sub-endothelial collagen matrix and platelet-surface receptor complex GPIb-IX-V. vWF also acts as a chaperone for coagulation factor VIII, delivering it to the site of injury, stabilizing its heterodimeric structure and protecting it from premature clearance from plasma. Mutations in VWF gene are linked to von Willebrand diseases 1-3 that are common hemorrhagic disorders caused by impaired platelet aggregation. Two isoforms of vWF have been described that are produced by alternative splicing.

Specificity

AB7356 is specific for vWF by ELISA against the human protein and by immunohistochemistry. The antibody is reactive in paraffin embedded tissues on human, rat, and mouse. Other species not tested. Antibody has not been evaluated by western blot analysis for reactivity to vWF.

Immunogen

Human von Willebrand Factor purified from plasma.

Application

Anti-von Willebrand Factor, Cat. No. AB7356, is a rabbit polyclonal antibody that detects von Willebrand Factor and has been tested for use in ELISA, Immunohistochemistry, and Western Blotting.
Immunohistochemistry Analysis: A representative lot detected von Willebrand Factor in Immunohistochemistry applications.

ELISA Analysis: A representative lot detected von Willebrand Factor in ELISA applications.

Immunohistochemistry Analysis: A 1:250-1,000 dilution from a representative lot detected von Willebrand Factor in human cerebral cortex, mouse hippocampus, rat cerebellum and human tonsil tissue sections.
Research Category
Cell Structure
Research Sub Category
ECM Proteins

Quality

Immunohistochemistry(paraffin):
Representative images from a previous lot.
Rabbit anti-vWF (AB7356) staining of Human Cerebral Cortex. Tissue pretreated with Citrate Buffer, ph 6.0, anti-vWF at 20 μg/mL, IHC-Select Detection with HRP-DAB.

Target description

250 kDa

Physical form

Format: Purified
Protein A purified
Purified rabbit immunoglobulin in buffer containing 0.02 M PBS pH 7.6, 0.25 M NaCl with 0.1% sodium azide.

Storage and Stability

Stable for 1 year at 2-8ºC from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Vein wall re-endothelialization after deep vein thrombosis is improved with low-molecular-weight heparin.
Moaveni, DK; Lynch, EM; Luke, C; Sood, V; Upchurch, GR; Wakefield, TW; Henke, PK
Journal of Vascular Surgery null
Antagonizing the ?v ?3 integrin inhibits angiogenesis and impairs woven but not lamellar bone formation induced by mechanical loading.
Tomlinson, RE; Schmieder, AH; Quirk, JD; Lanza, GM; Silva, MJ
Journal of Bone and Mineral Research null
Aleksander Skardal et al.
Stem cells translational medicine, 1(11), 792-802 (2012-12-01)
Stem cells obtained from amniotic fluid show high proliferative capacity in culture and multilineage differentiation potential. Because of the lack of significant immunogenicity and the ability of the amniotic fluid-derived stem (AFS) cells to modulate the inflammatory response, we investigated
Jason Baik et al.
PloS one, 7(10), e47494-e47494 (2012-10-17)
Clofazimine is a poorly-soluble but orally-bioavailable small molecule drug that massively accumulates in macrophages when administered over prolonged periods of time. To determine whether crystal-like drug inclusions (CLDIs) that form in subcellular spaces correspond to pure clofazimine crystals, macrophages of
Zhijie Wang et al.
Journal of applied physiology (Bethesda, Md. : 1985), 124(5), 1244-1253 (2018-01-26)
Pulmonary arterial hypertension (PAH) is a rapidly fatal disease in which mortality is due to right ventricular (RV) failure. It is unclear whether RV dysfunction initiates at the organ level or the subcellular level or both. We hypothesized that chronic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service