Skip to Content
Merck
All Photos(1)

Key Documents

AB3879

Sigma-Aldrich

Anti-Acetyl Lysine Antibody

Chemicon®, from rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity (predicted by homology)

all

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunofluorescence: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

shipped in

wet ice

target post-translational modification

acetylation (Lys)

General description

In the nucleus, DNA is tightly packed into nucleosomes generating an environment which is highly repressive towards DNA processes such as transcription. Acetylation of lysine residues within proteins has emerged as an important mechanism used by cells to overcome this repression. The acetylation of non-histone proteins such as transcription factors, as well as histones appears to be involved in this process. Acetylation may result in structural transitions as well as specific signaling within discrete chromatin domains. The role of acetylation in intracellular signaling has been inferred from the binding of acetylated peptides by the conserved bromodomain. Furthermore, recent findings suggest that bromodomain/acetylated-lysine recognition can serve as a regulatory mechanism in protein-protein interactions in numerous cellular processes such as chromatin remodeling and transcriptional activation.

Specificity

Recognizes proteins acetylated on lysine residues. This antibody recognizes multiple acetylated proteins including acetylated histones, acetylated BSA, and acetylated MBP, and exhibits no reaction with non-acetylated proteins.

Immunogen

Acetylated KLH.

Application

Anti-Acetyl Lysine Antibody is a high quality Rabbit Polyclonal Antibody for the detection of Acetyl Lysine & has been validated in ELISA, IF, IHC, IP & WB.
Immunoprecipitation:
A previous lot was used at 10 µg/mg crude protein.
ELISA:
0.25-1 μg/mL of a previous lot was used.

Immunohistochemistry:
2.5 μg/mL of a previous lot was used.

Immunofluorescence:
5 μg/mL of a previous lot was used.

Optimal working dilutions must be determined by end user.
Research Category
Signaling

Epigenetics & Nuclear Function
Research Sub Category
General Post-translation Modification

Histones

Quality

Routinely evaluated by Western Blot on Hela Acid Extract/Sodium Butyrate lysates.

Western Blot Analysis: 1:500 dilution of this lot detected lysine acylated proteins on 10 μg of Hela Acid Extract/Sodium Butyrate lysates.

Physical form

ImmunoAffinity Purified
Purified rabbit polyclonal in buffer containing PBS with 50% glycerol.

Storage and Stability

Stable for 1 year at -20°C in undiluted aliquots from date of receipt.

Handling Recommendations:
Upon first thaw, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance. Note: Variabillity in freezer temperatures below -20°C may cause glycerol containing solutions to become frozen during storage.

Analysis Note

Control
TSA-treated HeLa and NIH/3T3 cells, osteosarcoma tissue.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Niek G J Leus et al.
Biochemical pharmacology, 108, 58-74 (2016-03-20)
The increasing number of patients suffering from chronic obstructive pulmonary disease (COPD) represents a major and increasing health problem. Therefore, novel therapeutic approaches are needed. Class I HDACs 1, 2 and 3 play key roles in the regulation of inflammatory
Lorenzo Schiaffino et al.
Scientific reports, 8(1), 12875-12875 (2018-08-29)
Dysregulation in acetylation homeostasis has been implicated in the pathogenesis of the amyotrophic lateral sclerosis (ALS), a fatal neurodegenerative disorder. It is known that the acetylation of transcriptional factors regulates their activity. The acetylation state of NF-kB RelA has been
Terry R Medler et al.
Molecular cancer research : MCR, 14(10), 994-1008 (2016-07-01)
Stat5a is a transcription factor utilized by several cytokine/hormone receptor signaling pathways that promotes transcription of genes associated with proliferation, differentiation, and survival of cancer cells. However, there are currently no clinically approved therapies that directly target Stat5a, despite ample
Niek G J Leus et al.
Scientific reports, 7, 45047-45047 (2017-03-28)
Chronic obstructive pulmonary disease (COPD) constitutes a major health burden. Studying underlying molecular mechanisms could lead to new therapeutic targets. Macrophages are orchestrators of COPD, by releasing pro-inflammatory cytokines. This process relies on transcription factors such as NF-κB, among others.
Susanne Elfert et al.
PloS one, 8(4), e61867-e61867 (2013-04-25)
The members of the TCF/LEF family of DNA-binding proteins are components of diverse gene regulatory networks. As nuclear effectors of Wnt/β-catenin signaling they act as assembly platforms for multimeric transcription complexes that either repress or activate gene expression. Previously, it

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service