Skip to Content
Merck
All Photos(1)

Key Documents

AB1584

Sigma-Aldrich

Anti-c-Fos Antibody

Chemicon®, from sheep

Synonym(s):

G0/G1 Switch Regulatory Protein 7

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

sheep

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... FOS(2353)

General description

c-fos is a member of a family of immediate early gene (IEG) transcription factors (other members include Jun and Egr-1) also identified as proto-oncogenes. The basal expression of c-fos and other IEGs is typically low but increases relatively quickly and often dramatically (15-30 min for mRNA; 30-60 min for protein) in response to changes in cellular activity typically caused by external stimuli, such as stress or neuronal activation. Thus, these IEGs are being used to indicate specific activity driven cellular activation including the identification of neurons activated by various growth factors, neuroactive drugs, and correlated changes in behavioral or physiological states.

Specificity

Recognizes c-fos.

Immunogen

Synthetic c-fos peptide (residues Ser.Gly.Phe.Asn.Ala.Asp.Tyr.Glu.Ala. Ser.Ser.Ser.Arg.Cys) corresponding to amino acids 4-17 of human Fos, conjugated to bovine thyroglobulin.

Application

Immunohistochemistry: 1:500-1:4000, 2-8°C 1-3 days. Stains both fresh frozen and paraffin embedded tissue samples by indirect immunofluorescence and immunoperoxidase.

Recommended fixation is 4% PFA with 0.5% picric acid {Li et al 1999}; triton X-100 pretreatment in the block only; antigen retrieval for paraffin sections is recommended. Microwave citric acid treatment suggested.

Western blot: 10 μg/mL. Detects a band at approximately 55 kDa.

Optimal working dilutions must be determined by the end user.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors
Use Anti-c-Fos Antibody (Sheep Polyclonal Antibody) validated in IHC, IHC(P), WB to detect c-Fos also known as G0/G1 Switch Regulatory Protein 7.

Target description

55 kDa

Physical form

Ammonium Sulfate Precipitation
Format: Purified
Purified immunoglobulin. Lyophilized, no preservatives. Reconstitute with 500 μL of sterile distilled water (1 mg/mL).

Storage and Stability

Maintain lyophilized material at -20 to -70°C for up to 12 months. After reconstitution maintain at -20°C in undiluted aliquots for up to one year.

Avoid repeated freeze/thaw cycles.

During shipment, small volumes of product will occasionally become entrapped in the seal of the product vial. For products with volumes of 200μL or less, we recommend gently tapping the vial on a hard surface or briefly centrifuging the vial in a tabletop centrifuge to dislodge any liquid in the container′s cap.

Analysis Note

Control
Widely expressed

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Min Jee Kim et al.
Molecular pain, 14, 1744806918814345-1744806918814345 (2018-11-02)
The insular cortex is an important region of brain involved in the processing of pain and emotion. Recent studies indicate that lesions in the insular cortex induce pain asymbolia and reverse neuropathic pain. Endogenous cannabinoids (endocannabinoids), which have been shown
V Kafetzopoulos et al.
Molecular psychiatry, 23(3), 579-586 (2017-04-12)
The hippocampus and prefrontal cortex (PFC) are connected in a reciprocal manner: whereas the hippocampus projects directly to the PFC, a polysynaptic pathway that passes through the nucleus reuniens (RE) of the thalamus relays inputs from the PFC to the
Anna Woodbury et al.
Journal of natural products, 78(11), 2531-2536 (2015-11-06)
This study examined the short- and long-term neuroprotective and analgesic activity of honokiol (a naturally occurring lignan isolated from Magnolia) on developing brains in neonates exposed to inflammatory pain, known to cause neuronal cell death. Postnatal day 4 (P4) neonatal
Maria Scherma et al.
Nutrients, 15(17) (2023-09-09)
Anorexia nervosa (AN) is a complex eating disorder characterized by reduced caloric intake to achieve body-weight loss. Furthermore, over-exercise is commonly reported. In recent years, animal models of AN have provided evidence for neuroplasticity changes in specific brain areas of
Yoko Kato et al.
Scientific reports, 11(1), 3348-3348 (2021-02-10)
Multiple sequential actions, performed during parental behaviors, are essential elements of reproduction in mammalian species. We showed that neurons expressing melanin concentrating hormone (MCH) in the lateral hypothalamic area (LHA) are more active in rodents of both sexes when exhibiting

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service