Skip to Content
Merck
All Photos(3)

Key Documents

AB1543

Sigma-Aldrich

Anti-Synapsin I Antibody

serum, Chemicon®

Synonym(s):

Synapsin-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, bovine, rat, mouse

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable (paraffin)
immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... SYN1(6853)

General description

Synapsin I is a member of the synapsin family. Synapsins are neuronal phosphoproteins which associate with the cytoplasmic surface of synaptic vesicles. Family members are characterized by common protein domains, and they are implicated in synaptogenesis and the modulation of neurotransmitter release, suggesting a potential role in several neuropsychiatric diseases. This member of the synapsin family plays a role in regulation of axonogenesis and synaptogenesis. The protein serves as a substrate for several different protein kinases and phosphorylation may function in the regulation of this protein in the nerve terminal. Mutations of the Synapsin I gene may be associated with X linked disorders with primary neuronal degeneration such as Rett syndrome.

Specificity

Synapsin I (Mr of 80,000 and 77,000). Immunolabeling is blocked by preadsorption of antibody with Synapsin I. Synapsin is located only in brain nerve terminal and is an excellent marker for synapses.

Immunogen

Synapsin I (mixture of Ia & Ib) purified from bovine brain.

Application

Detect Synapsin I using this Anti-Synapsin I Antibody validated for use in IC, IH(P), IP & WB.
Research Category
Neuroscience
Research Sub Category
Synapse & Synaptic Biology
Western blotting:
1:200 of a previous lot was used for labeled Protein A, 1:1,000 dilution for ECL.

Immunocytochemistry:
1:500-1:2,000 dilution of a previous lot was used.

Immunoprecipitation:
5 μL of a previous lot quantitatively immunoprecipitated all synapsin in 200 μg of rat brain.

Optimal working dilutions must be determined by the end user.

Quality

Immunohistochemistry(paraffin): Synapsin representative staining pattern/morphology in rat hippocampal neurons. Tissue was pretreated with Citrate pH 6.0, antigen retrieval. A previous lot of this antibody was diluted to 1:1000, using IHC-Select reagents used with HRP-DAB. Immunoreactivity is seen directly associated with terminal end of neuronal cell body.
IHC-Paraffin Staining With Epitope Retrieval: Rat Hippocampus

Target description

77 & 80 kDa

Physical form

Rabbit polyclonal antiserum in buffer containing no preservatives.
Unpurified

Storage and Stability

Stable for 1 year at -20ºC from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
Mouse hippocampal primary neurons, Mouse brain lysates, PC-12 cells.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael J Vasek et al.
Nature, 534(7608), 538-543 (2016-06-24)
Over 50% of patients who survive neuroinvasive infection with West Nile virus (WNV) exhibit chronic cognitive sequelae. Although thousands of cases of WNV-mediated memory dysfunction accrue annually, the mechanisms responsible for these impairments are unknown. The classical complement cascade, a
Innate synchronous oscillations in freely-organized small neuronal circuits.
Shein Idelson, M; Ben-Jacob, E; Hanein, Y
Testing null
Fernando J Sepulveda et al.
PloS one, 5(7), e11820-e11820 (2010-08-03)
The mechanisms that induce Alzheimer's disease (AD) are largely unknown thereby deterring the development of disease-modifying therapies. One working hypothesis of AD is that Abeta excess disrupts membranes causing pore formation leading to alterations in ionic homeostasis. However, it is
Franziska Neuser et al.
PloS one, 8(6), e66497-e66497 (2013-07-11)
Enduring reorganization is accepted as a fundamental process of adult neural plasticity. The most dramatic example of this reorganization is the birth and continuously occurring incorporation of new neurons into the pre-existing network of the adult mammalian hippocampus. Based on
The X-linked mental retardation protein OPHN1 interacts with Homer1b/c to control spine endocytic zone positioning and expression of synaptic potentiation.
Nakano-Kobayashi, A; Tai, Y; Nadif Kasri, N; Van Aelst, L
The Journal of Neuroscience null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service