Skip to Content
Merck
All Photos(1)

Key Documents

8.57008

Sigma-Aldrich

Oxidizing Reagent for DNA Synthesis

0.05M Iodine in Pyridine, Novabiochem®

Synonym(s):

0.05M Iodine in Pyridine, Novabiochem®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352401
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

200
300

product line

Novabiochem®

form

liquid

reaction suitability

reaction type: Oxidations
reagent type: oxidant

manufacturer/tradename

Novabiochem®

transition temp

flash point 20.0 °C

density

0.99 g/cm3

storage temp.

2-30°C

General description

Pre-mixed ready-to-use Deblock Reagent
  • 857008.1000: Compatible with Cytiva ÄKTA DNA Synthesisers. Stand alone from Instrument (GL-45 Neck Finish)
  • 857008.2500: Compatible with Cytiva ÄKTA DNA Synthesisers. Stand alone from Instrument (GL-45 Neck Finish)
  • 857008.4000: Compatible with MerMade DNA Synthesisers. Stand alone from Instrument (38mm Neck Finish)

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT RE 2 Oral

Target Organs

Thyroid

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

68.0 °F

Flash Point(C)

20.0 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service