Skip to Content
Merck
All Photos(1)

Key Documents

8.55084

Sigma-Aldrich

Aminomethyl NovaGel

Novabiochem®

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
13111023
NACRES:
NA.22

product line

NovaGel
Novabiochem®

Quality Level

form

beads

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

amine

storage temp.

2-8°C

General description

Aminomethyl NovaGel is a PEG-PS resin which has been designed to meet the requirements of organic chemists for resins of high substitution coupled with broad solvent compatibility. This support has a nominal loading of 0.7 mmole/g – almost twice that of comparable resins – but still retains most of the swelling characteristics of conventional PEG-PS supports. It is prepared from a special high-swell version of AM resin by partial derivatization with methyl-PEG-p-nitrophenylcarbonate [1]. Resin functionality is provided by residual aminomethyl groups. The urethane linkage between the PEG and the base resin is stable to strongly acidic, basic and reducing conditions, ensuring minimal loss of PEG chains during synthesis. Furthermore, because the linker is not attached to the end of the PEG chains, if leaching of PEG does occur, the product is not contaminated with U.V. absorbing linker-PEG fragments which can complicate the nmr spectra and HPLC profiles of products obtained with other PEG-PS supports.

Associated Protocols and Technical Articles
Protocols for Loading of Peptide Synthesis Resins

Literature references

[1] J. H. Adams, et al. (1998) J. Org. Chem., 63, 3706.

Linkage

Replaces: 01-64-0283

Analysis Note

Color (visual): white to yellow to beige
Appearance of substance (visual): beads
Loading (determined from the substitution of the Fmoc-Leu loaded resin): 0.60 - 0.90 mmol/g
TNBS-Test: passes test
Swelling Volume (in DMF): lot specific result
The PEG-PS resin is prepared from aminomethyl copoly (Styrene 1% DVB), 100 - 200 mesh.

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Novabiochem® offers polymer supports for solid phase peptide synthesis, suitable for various peptide lengths and sequences.

Novabiochem® offers polymer supports for solid phase peptide synthesis, suitable for various peptide lengths and sequences.

Novabiochem® offers polymer supports for solid phase peptide synthesis, suitable for various peptide lengths and sequences.

Novabiochem® offers polymer supports for solid phase peptide synthesis, suitable for various peptide lengths and sequences.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service