8.52021
Fmoc-Val-OH
Novabiochem®
Synonym(s):
Fmoc-Val-OH, N-α-Fmoc-L-valine
About This Item
Quality Level
product line
Novabiochem®
Assay
≥98% (TLC)
≥98.0% (acidimetric)
≥99.0% (HPLC)
form
powder
reaction suitability
reaction type: Fmoc solid-phase peptide synthesis
manufacturer/tradename
Novabiochem®
mp
140-145 °C
application(s)
peptide synthesis
functional group
Fmoc
storage temp.
2-30°C
SMILES string
N(C(C(C)C)C(=O)O)C(=O)OCC1c2c(cccc2)c3c1cccc3
InChI
1S/C20H21NO4/c1-12(2)18(19(22)23)21-20(24)25-11-17-15-9-5-3-7-13(15)14-8-4-6-10-16(14)17/h3-10,12,17-18H,11H2,1-2H3,(H,21,24)(H,22,23)
InChI key
UGNIYGNGCNXHTR-UHFFFAOYSA-N
General description
Standard building block for introduction of valine amino-acid residues by Fmoc SPPS
Associated Protocols and Technical Articles
Fmoc-amino acids for Peptide Production
Cleavage and Deprotection Protocols for Fmoc SPPS
Application
- Evaluation of Met-Val-Lys as a Renal Brush Border Enzyme-Cleavable Linker: This study explores using Fmoc-Val-OH in the development of renal-targeted drug delivery systems for enhanced therapeutic efficacy (Bendre et al., 2020).
- Enantioselective Ortho-CH Cross-Coupling of Diarylmethylamines with Organoborons: Reports the use of Fmoc-Val-OH in a new methodology for achieving high yields and enantioselectivity in cross-coupling reactions (Laforteza et al., 2015).
- AJIPHASE®: A Highly Efficient Synthetic Method for One-Pot Peptide Elongation: Demonstrates a novel approach using Fmoc-Val-OH for the scalable synthesis of peptides, showing significant improvements in efficiency and scalability (Takahashi et al., 2017).
Linkage
Analysis Note
Appearance of substance (visual): powder
Colour index (0,5 M in DMF): ≤ 150 Hazen
Identity (IR): passes test
Enantiomeric purity: ≥ 99.8 % (a/a)
Purity (HPLC): ≥ 99.0 % (a/a)
Fmoc-ß-Ala-OH (HPLC): ≤ 0.1 % (a/a)
Fmoc-ß-Ala-Val-OH (HPLC): ≤ 0.1 % (a/a)
Fmoc-Val-Val-OH (HPLC): ≤ 0.1 % (a/a)
Assay free amino acid (GC): ≤ 0.2 %
Purity (TLC(011A)): ≥ 98 %
Purity (TLC(0811)): ≥ 98 %
Solubility (25 mmole in 50 ml DMF): clearly soluble
Assay (acidimetric): ≥ 98.0 %
Water (K. F.): ≤ 2.0 %
Ethyl acetate (HS-GC): ≤ 0.5 %
Acetate (IC): ≤ 0.02 %
To see the solvent systems used for TLC of Novabiochem® products please click here.
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service