Skip to Content
Merck
All Photos(2)

Key Documents

8.22259

Sigma-Aldrich

Benzyl alcohol

for synthesis

Synonym(s):

Benzenemethanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2OH
CAS Number:
Molecular Weight:
108.14
Beilstein:
878307
MDL number:
UNSPSC Code:
12352104
EC Index Number:
202-859-9
NACRES:
NA.22

vapor density

3.7 (vs air)

Quality Level

vapor pressure

0.07 hPa ( 20 °C)
13.3 mmHg ( 100 °C)
3.75 mmHg ( 77 °C)

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

435 °C (DIN 51794)
817 °F

potency

1620 mg/kg LD50, oral (Rat)

refractive index

n20/D 1.539 (lit.)

bp

203-205 °C (lit.)

mp

−16-−13 °C (lit.)

transition temp

flash point 101 °C (DIN 51758)

density

1.045 g/mL at 25 °C (lit.)

storage temp.

2-30°C

SMILES string

OCc1ccccc1

InChI

1S/C7H8O/c8-6-7-4-2-1-3-5-7/h1-5,8H,6H2

InChI key

WVDDGKGOMKODPV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Heterojunction photocatalysts for hydrogen evolution and benzyl alcohol oxidation: Wang et al. developed phosphotungstic acid clusters on Znln2S4 nanoflowers, demonstrating their efficiency as molecular-scale S-scheme heterojunctions for simultaneous hydrogen evolution and benzyl alcohol oxidation, indicating a promising application in renewable energy and organic synthesis technologies (Wang et al., 2024).
  • Photoelectrocatalytic alcohol oxidation: Gong et al. enhanced the performance of a BiVO(4) photoanode in bicarbonate electrolyte for efficient photoelectrocatalytic oxidation of alcohols including benzyl alcohol, showing potential for clean energy production and organic pollutant degradation (Gong et al., 2024).

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.045 - 1.046
Water (K. F.): ≤ 0.10 %
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

213.8 °F - DIN 51758

Flash Point(C)

101 °C - DIN 51758


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service