Skip to Content
Merck
All Photos(1)

Key Documents

8.21178

Sigma-Aldrich

Trimethylammonium chloride

for synthesis

Synonym(s):

Trimethylammonium chloride, Trimethylamine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3N · HCl
CAS Number:
Molecular Weight:
95.57
MDL number:
UNSPSC Code:
12352107
EC Index Number:
209-810-0
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

form

crystals

potency

3090 mg/kg LD50, oral (Rat)

pH

5 (20 °C, 100 g/L in H2O)

mp

273-278 °C

storage temp.

2-30°C

SMILES string

[Cl-].[N+H](C)(C)C

InChI

1S/C3H9N.ClH/c1-4(2)3;/h1-3H3;1H

InChI key

SZYJELPVAFJOGJ-UHFFFAOYSA-N

Analysis Note

Assay (argentometric): ≥ 98.0 %
Melting range (lower value): ≥ 273 °C
Melting range (upper value): ≤ 278 °C
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service