8.20708
2-Hydroxyethanesulfonic acid sodium salt
for synthesis
Synonym(s):
2-Hydroxyethanesulfonic acid sodium salt, Isethionic acid sodium salt, Sodium isethionate
About This Item
Quality Level
form
powder
potency
>2500 mg/kg LD50, oral (Rat)
pH
7.0-11.0 (20 °C, 20 g/L in H2O)
mp
192-194 °C
bulk density
800 kg/m3
storage temp.
2-30°C
SMILES string
[Na+].[S](=O)(=O)([O-])CCO
InChI
1S/C2H6O4S.Na/c3-1-2-7(4,5)6;/h3H,1-2H2,(H,4,5,6);/q;+1/p-1
InChI key
LADXKQRVAFSPTR-UHFFFAOYSA-M
Application
- Investigation on specific heat capacity and thermal behavior of sodium hydroxyethyl sulfonate: This study presents a detailed analysis of the thermal behavior and specific heat capacity of sodium hydroxyethyl sulfonate, also known as 2-hydroxyethanesulfonic acid sodium salt (SHES) (H Hao, Y Zhang, X Chen, 2017).
- Design and Mechanism of a Self‐Powered and Disintegration–Reorganization–Regeneration Power Supply with Cold Resistance: The research explores a novel power supply system using 2-hydroxyethanesulfonic acid sodium salt, focusing on its cold resistance characteristics (H Dai, Y Chen, W Dai, Z Hu, M Li, W Zhang, 2021).
Analysis Note
Identity (IR): passes test
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service