Skip to Content
Merck
All Photos(1)

Key Documents

8.20234

Sigma-Aldrich

Benzaldehyde oxime

for synthesis

Synonym(s):

Benzaldehyde oxime, Benzaldoxime

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH=NOH
CAS Number:
Molecular Weight:
121.14
MDL number:
UNSPSC Code:
12352126
EC Index Number:
213-261-2
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

≥96.5% (GC)

form

crystals

bp

123 °C/1013 hPa

mp

30 °C

transition temp

flash point 108 °C

density

1.11 g/cm3

storage temp.

2-30°C

InChI

1S/C7H7NO/c9-8-6-7-4-2-1-3-5-7/h1-6,9H/b8-6+

InChI key

VTWKXBJHBHYJBI-SOFGYWHQSA-N

Analysis Note

Assay (GC, area%): ≥ 96.5 % (a/a)
Melting range (lower value): ≥ 24 °C
Melting range (upper value): ≤ 27 °C
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service