Skip to Content
Merck
All Photos(1)

Key Documents

8.19015

Sigma-Aldrich

Polyethylene glycol

3000, for synthesis

Synonym(s):

Poly(ethylene glycol), Polyethylene glycol 3000, Polyglycol, Polyethylene oxide, Polyoxy ethylene, PEG 3000, PEG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H(OCH2CH2)nOH
CAS Number:
MDL number:
UNSPSC Code:
13111021
EC Index Number:
500-038-2
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

<0.1 hPa ( 20 °C)

Quality Level

form

liquid

autoignition temp.

420 °C

potency

28000 mg/kg LD50, oral (Rat)
>20000 mg/kg LD50, skin (Rabbit)

pH

4-7 (20 °C, 100 g/L in H2O)

mp

55-58 °C

transition temp

flash point 270 °C

density

1.21 g/cm3 at 20 °C

bulk density

400‑500 kg/m3

storage temp.

2-30°C

SMILES string

C(CO)O

InChI

1S/C2H6O2/c3-1-2-4/h3-4H,1-2H2

InChI key

LYCAIKOWRPUZTN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Polyethylene glycol 3000 can be used as:
  • A surface-modification agent in the synthesis of monodispersed magnetite colloidal magnetic nanoparticles.[1]
  • A precipitating agent in the crystallization of oligoxyloglucan reducing-end specific cellobiohydrolase (OXG-RCBH) by the hanging-drop vapor diffusion method.[2]
  • A solvent and phase-transfer catalyst (PTC) in the application of aqueous biphasic reactive extraction.[3]
  • An additive in the preparation of polysulfone membrane to study the porometry.[4]

Analysis Note

Hydroxyl value: 34 - 42
Melting range (lower value): ≥ 55 °C
Melting range (upper value): ≤ 59 °C
Average molecular mass: 2700 - 3300
Identity (IR): passes test

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

281.5 °F - closed cup

Flash Point(C)

138.6 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Porometry studies of the polysulfone membranes on addition of poly (ethylene glycol) in gelation bath during preparation
Yogesh SJ, et al.
Journal of the Mexican Chemical Society, 52(2) (2008)
Katsuro Yaoi et al.
Acta crystallographica. Section D, Biological crystallography, 59(Pt 10), 1838-1839 (2003-09-23)
A novel xyloglucan-specific exo-beta-glycosidase, oligoxyloglucan reducing-end specific cellobiohydrolase (OXG-RCBH), recognizes the reducing end of oligoxyloglucan and releases two glucosyl residue segments from the main chain. OXG-RCBH was crystallized by the hanging-drop vapour-diffusion method with polyethylene glycol 3000 and polyethylene glycol
Eleftherios Halevas et al.
Journal of inorganic biochemistry, 213, 111271-111271 (2020-10-19)
Targeted tissue drug delivery is a challenge in contemporary nanotechnologically driven therapeutic approaches, with the interplay interactions between nanohost and encapsulated drug shaping the ultimate properties of transport, release and efficacy of the drug at its destination. Prompted by the
Polyethylene glycol and solutions of polyethylene glycol as green reaction media
Chen Ji, et al.
Green Chemistry, 7(2) (2005)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service