Skip to Content
Merck
All Photos(1)

Key Documents

8.18035

Sigma-Aldrich

Methyl trifluoromethanesulfonate

for synthesis

Synonym(s):

Methyl trifluoromethanesulfonate, Trifluoromethanesulfonic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H3F3O3S
CAS Number:
Molecular Weight:
164.10
MDL number:
UNSPSC Code:
12352108
EC Index Number:
206-371-7
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

form

liquid

bp

99 °C/1013 hPa

transition temp

flash point 38 °C

density

1.49 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

FC(F)(F)[S](=O)(=O)OC

InChI

1S/C2H3F3O3S/c1-8-9(6,7)2(3,4)5/h1H3

InChI key

OIRDBPQYVWXNSJ-UHFFFAOYSA-N

Analysis Note

Assay 1H-NMR: ≥ 90.0 %
Purity 19F-NMR: ≥ 98.0 %
Density (d 20 °C/ 4 °C): 1.490 - 1.500
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

100.4 °F - closed cup

Flash Point(C)

38 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service