Skip to Content
Merck
All Photos(1)

Key Documents

8.14100

Sigma-Aldrich

3-Diethylamino-1-propylamine

for synthesis

Synonym(s):

3-Diethylamino-1-propylamine, N,N-Diethyl-1,3-diaminopropane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NCH2CH2CH2N(C2H5)2
CAS Number:
Molecular Weight:
130.23
MDL number:
UNSPSC Code:
12352116
EC Index Number:
203-236-4
NACRES:
NA.22

vapor pressure

2.2 hPa ( 20 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

autoignition temp.

215 °C

potency

410 mg/kg LD50, oral (Rat)

expl. lim.

1.0 % (v/v)

pH

11.7 (20 °C, 10 g/L in H2O)

bp

168-171 °C

mp

-60 °C

transition temp

flash point 51 °C

density

0.82 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

N(CCCN)(CC)CC

InChI

1S/C7H18N2/c1-3-9(4-2)7-5-6-8/h3-8H2,1-2H3

InChI key

QOHMWDJIBGVPIF-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.824 - 0.827
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1A - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

125.6 °F - closed cup

Flash Point(C)

52 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service