Skip to Content
Merck
All Photos(1)

Key Documents

8.07466

Sigma-Aldrich

2-Pyridinecarbaldehyde oxime

for synthesis

Synonym(s):

2-Pyridinecarbaldehyde oxime, Picoline aldoxime

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
2-(CH=NOH)C5H4N
CAS Number:
Molecular Weight:
122.12
MDL number:
UNSPSC Code:
12352126
EC Index Number:
212-849-6
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

form

crystals

potency

388 mg/kg LD50, oral (Rat)

pH

7 (20 °C, 13 g/L in H2O)

mp

110-112 °C

solubility

15 g/L

bulk density

400‑500 kg/m3

storage temp.

2-30°C

SMILES string

N1\C(=C\N=O)\C=CC=C1

InChI

1S/C6H6N2O/c9-8-5-6-3-1-2-4-7-6/h1-5,7H/b6-5+

InChI key

VRGKDCRTYBCJTA-AATRIKPKSA-N

Analysis Note

Assay (HClO₄): ≥ 99.0 %
Melting range (lower value): ≥ 110 °C
Melting range (upper value): ≤ 112 °C
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service