Skip to Content
Merck
All Photos(1)

Key Documents

8.07049

Sigma-Aldrich

4-Methylpyridine

for synthesis

Synonym(s):

4-Methylpyridine, 4-Picoline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H7N
CAS Number:
Molecular Weight:
93.13
MDL number:
UNSPSC Code:
12352005
EC Index Number:
203-626-4
NACRES:
NA.22

vapor pressure

5.8 hPa ( 20 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

autoignition temp.

>500 °C

potency

1290 mg/kg LD50, oral (Rat)
270 mg/kg LD50, skin (Rabbit)

expl. lim.

1.3-8.7 % (v/v)

bp

145 °C/1013 hPa

mp

3 °C

transition temp

flash point 40 °C

solubility

1000 g/L

density

0.95 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

n1ccc(cc1)C

InChI

1S/C6H7N/c1-6-2-4-7-5-3-6/h2-5H,1H3

InChI key

FKNQCJSGGFJEIZ-UHFFFAOYSA-N

Application

<ul>
<li><strong>Hemocompatible nanomaterials for biosensing: </strong>4-Methylpyridine is used as the functionalizing agent in the synthesis of the 3-n-propyl(2-amino-4-methyl)pyridium chloride ( SiAMPy+Cl-) material through a sol-gel procedure for the production of stable nanoconjugates with gold nanoparticles (AuNPs-SiAMPy+) (Lima et al., 2022).</li>
</ul>

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.953 - 0.955
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

102.2 °F - closed cup

Flash Point(C)

39 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service