Skip to Content
Merck
All Photos(1)

Key Documents

8.01811

Sigma-Aldrich

Benzyl cyanide

for synthesis

Synonym(s):

Benzyl cyanide, Phenylacetonitrile, α-Tolunitrile

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H7N
CAS Number:
Molecular Weight:
117.15
MDL number:
UNSPSC Code:
12352117
EC Index Number:
205-410-5
form:
liquid
Assay:
≥99.0% (GC)

vapor pressure

0.2 hPa ( 20 °C)

Quality Level

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

590 °C

potency

270 mg/kg LD50, oral (Rat)
270 mg/kg LD50, skin (Rabbit)

bp

66 °C/1013 hPa

mp

-24 °C

transition temp

flash point 102 °C

solubility

0.1 g/L

density

1.02 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C8H7N/c9-7-6-8-4-2-1-3-5-8/h1-5H,6H2

InChI key

SUSQOBVLVYHIEX-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.015 - 1.017
Identity (IR): passes test

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

215.6 °F - closed cup

Flash Point(C)

102 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service