Skip to Content
Merck
All Photos(1)

Key Documents

8.01808

Sigma-Aldrich

Ethyl benzoylacetate

for synthesis

Synonym(s):

Ethyl benzoylacetate, Benzoylacetic acid ethyl ester, 3-Oxo-3-phenylpropionic acid ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H12O3
CAS Number:
Molecular Weight:
192.21
MDL number:
UNSPSC Code:
12352108
EC Index Number:
202-295-3
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

1 hPa ( 108 °C)

Quality Level

Assay

≥95.0% (acidimetric)

form

liquid

autoignition temp.

375 °C (DIN 51794)

potency

>5000 mg/kg LD50, oral (Rat)

transition temp

flash point 140 °C

density

1.114 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O(CC)C(=O)CC(=O)c1ccccc1

InChI

1S/C11H12O3/c1-2-14-11(13)8-10(12)9-6-4-3-5-7-9/h3-7H,2,8H2,1H3

InChI key

GKKZMYDNDDMXSE-UHFFFAOYSA-N

Analysis Note

Assay (acidimetric): ≥ 95.0 %
Density (d 20 °C/ 4 °C): 1.105 - 1.120
Identity (IR): passes test

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

284.0 °F - closed cup

Flash Point(C)

140 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service