Skip to Content
Merck
All Photos(1)

Key Documents

8.00741

Sigma-Aldrich

Tetrachlorophthalic anhydride

for synthesis

Synonym(s):

Tetrachlorophthalic anhydride, Tetrachloro-1,2-benzenedicarboxylic anhydride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8Cl4O3
CAS Number:
Molecular Weight:
285.90
MDL number:
UNSPSC Code:
12352115
EC Index Number:
204-171-4
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

0.0021 kPa ( 145 °C)

Quality Level

Assay

≥99.0% (GC)

form

powder

potency

>15800 mg/kg LD50, oral (Rat)
>5000 mg/kg LD50, skin (Rabbit)

bp

350 °C/1.013 hPa

mp

253-256 °C

storage temp.

2-30°C

SMILES string

Clc1c2c(c(c(c1Cl)Cl)Cl)C(=O)OC2=O

InChI

1S/C8Cl4O3/c9-3-1-2(8(14)15-7(1)13)4(10)6(12)5(3)11

InChI key

AUHHYELHRWCWEZ-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Melting range (lower value): ≥ 253 °C
Melting range (upper value): ≤ 256 °C
Hexachlorobenzene: ≤ 0.10 % (a/a)
Identity (IR): passes test

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

680.0 °F - closed cup

Flash Point(C)

360 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service