Skip to Content
Merck
All Photos(1)

Key Documents

8.00468

Sigma-Aldrich

Benzenesulfonic acid

for synthesis

Synonym(s):

Benzenesulfonic acid, Phenylsulfonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6O3S
CAS Number:
Molecular Weight:
158.18
MDL number:
UNSPSC Code:
12352106
EC Index Number:
202-638-7
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

≥97.0% (acidimetric)

form

solid

potency

1175 mg/kg LD50, oral (Rat)

pH

2 (20 °C in H2O, saturated solution)

bp

171 °C/0.13 hPa

mp

43 °C

transition temp

flash point >100 °C

solubility

930 g/L

density

1.32 g/cm3 at 47 °C

storage temp.

2-30°C

SMILES string

[S](=O)(=O)([O-])c1ccccc1.[H+]

InChI

1S/C6H6O3S/c7-10(8,9)6-4-2-1-3-5-6/h1-5H,(H,7,8,9)

InChI key

SRSXLGNVWSONIS-UHFFFAOYSA-N

Analysis Note

Assay (acidimetric): ≥ 97.0 %
Water (K. F.): ≤ 2.3 %
Sulfuric acid: ≤ 0.7 %
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

212.0 °F

Flash Point(C)

> 100 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service