Skip to Content
Merck
All Photos(1)

Key Documents

8.00399

Sigma-Aldrich

Myristic acid

for synthesis

Synonym(s):

Myristic acid, Tetradecanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)12COOH
CAS Number:
Molecular Weight:
228.37
MDL number:
UNSPSC Code:
12352106
EC Index Number:
208-875-2
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

<0.01 hPa ( 20 °C)

Quality Level

Assay

≥99.0% (GC)

form

solid

potency

>10000 mg/kg LD50, oral (Rat)

bp

326 °C/1013 hPa

mp

52-56 °C

transition temp

flash point >165 °C

solubility

1.07 mg/L

density

0.8622 g/cm3 at 54 °C

storage temp.

2-30°C

SMILES string

OC(=O)CCCCCCCCCCCCC

InChI

1S/C14H28O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14(15)16/h2-13H2,1H3,(H,15,16)

InChI key

TUNFSRHWOTWDNC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Melting range (lower value): ≥ 52 °C
Melting range (upper value): ≤ 56 °C
Identity (IR): passes test

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service